Assay Details
Gene Symbol: B4GALT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_004775.3 | NP_004766.2 | 1 - 2 | 412 | 81 | |
XM_006722578.3 | XP_006722641.1 | 1 - 2 | 1380 | 81 | ||
GenBank mRNA | AB024742.2 | - | 1 - 2 | 121 | 81 | |
AF038664.1 | - | 1 - 2 | 390 | 81 | ||
AF097159.1 | - | 1 - 2 | 412 | 81 | ||
AK314860.1 | - | 1 - 2 | 270 | 81 | ||
BC069620.1 | - | 1 - 2 | 193 | 7262397 | 81 | |
BC069642.1 | - | 1 - 2 | 192 | 7262468 | 81 | |
BC074835.2 | - | 1 - 2 | 133 | 30915327 | 81 | |
BC074884.2 | - | 1 - 2 | 133 | 30915473 | 81 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC18000440.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR18004494.hg.1 | ENST00000306851.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR18004494.hg.1 | ENST00000383131.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR18004494.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR18004494.hg.1 | TC18000440.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR18004500.hg.1 | ENST00000237019.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR18004500.hg.1 | ENST00000306851.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR18004500.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR18004500.hg.1 | TC18000440.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR18004501.hg.1 | ENST00000306851.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR18004501.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR18004501.hg.1 | TC18000440.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000237019.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000237019.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000237019.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000237019.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000237019.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000237019.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000237019.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000237019.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000237019.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000237019.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000237019.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000237019.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000237019.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000306851.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000306851.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000306851.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000306851.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000306851.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000306851.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000306851.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000306851.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000306851.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000306851.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000306851.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000306851.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000306851.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000306851.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000306851.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000306851.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000383131.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000383131.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000383131.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000383131.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000383131.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000383131.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000383131.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000383131.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000383131.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000383131.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000383131.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000578114.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000578114.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000578114.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000578114.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000578114.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000578114.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000578114.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000578114.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000578114.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000578114.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000578114.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000579372.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000579372.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000579372.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR18004497.hg.1 | ENST00000237019.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR18004497.hg.1 | ENST00000306851.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR18004497.hg.1 | ENST00000383131.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR18004497.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR18004497.hg.1 | TC18000440.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR18004498.hg.1 | ENST00000237019.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR18004498.hg.1 | ENST00000306851.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR18004498.hg.1 | ENST00000383131.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR18004498.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR18004498.hg.1 | TC18000440.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR18004495.hg.1 | ENST00000237019.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR18004495.hg.1 | ENST00000306851.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR18004495.hg.1 | ENST00000383131.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR18004495.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR18004495.hg.1 | TC18000440.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1800157240.hg.1 | ENST00000237019.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800157240.hg.1 | ENST00000306851.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800157240.hg.1 | ENST00000383131.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800157240.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800157240.hg.1 | TC1800008399.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1800157240.hg.1 | ENST00000237019.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800157240.hg.1 | ENST00000306851.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800157240.hg.1 | ENST00000383131.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800157240.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800157239.hg.1 | ENST00000306851.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800157239.hg.1 | ENST00000383131.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800157239.hg.1 | ENST00000579372.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800157239.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800157239.hg.1 | TC1800008399.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1800157239.hg.1 | ENST00000306851.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800157239.hg.1 | ENST00000383131.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800157239.hg.1 | ENST00000579372.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800157239.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800157242.hg.1 | ENST00000237019.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800157242.hg.1 | ENST00000306851.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800157242.hg.1 | ENST00000383131.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800157242.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800157242.hg.1 | TC1800008399.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1800157242.hg.1 | ENST00000237019.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800157242.hg.1 | ENST00000306851.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800157242.hg.1 | ENST00000383131.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800157242.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800157244.hg.1 | ENST00000237019.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800157244.hg.1 | ENST00000306851.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800157244.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800157244.hg.1 | TC1800008399.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1800157244.hg.1 | ENST00000237019.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800157244.hg.1 | ENST00000306851.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800157244.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800157245.hg.1 | ENST00000306851.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800157245.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800157245.hg.1 | TC1800008399.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1800157245.hg.1 | ENST00000306851.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800157245.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000237019.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000237019.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000237019.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000237019.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000237019.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000237019.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000237019.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000237019.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000237019.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000237019.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000237019.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000237019.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000237019.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000306851.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000306851.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000306851.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000306851.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000306851.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000306851.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000306851.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000306851.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000306851.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000306851.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000306851.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000306851.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000306851.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000306851.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000306851.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000306851.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000383131.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000383131.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000383131.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000383131.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000383131.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000383131.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000383131.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000383131.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000383131.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000383131.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000383131.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000578114.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000578114.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000578114.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000578114.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000578114.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000578114.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000578114.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000578114.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000578114.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000578114.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000578114.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000579372.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000579372.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000579372.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | NM_004775.3 | Refseq |
