Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008399.hg.1 |
ENST00000237019.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008399.hg.1 |
ENST00000237019.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008399.hg.1 |
ENST00000237019.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008399.hg.1 |
ENST00000237019.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008399.hg.1 |
ENST00000237019.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008399.hg.1 |
ENST00000237019.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008399.hg.1 |
ENST00000237019.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008399.hg.1 |
ENST00000237019.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008399.hg.1 |
ENST00000237019.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008399.hg.1 |
ENST00000237019.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008399.hg.1 |
ENST00000237019.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008399.hg.1 |
ENST00000237019.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008399.hg.1 |
ENST00000237019.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008399.hg.1 |
ENST00000306851.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008399.hg.1 |
ENST00000306851.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008399.hg.1 |
ENST00000306851.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008399.hg.1 |
ENST00000306851.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008399.hg.1 |
ENST00000306851.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008399.hg.1 |
ENST00000306851.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008399.hg.1 |
ENST00000306851.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008399.hg.1 |
ENST00000306851.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008399.hg.1 |
ENST00000306851.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008399.hg.1 |
ENST00000306851.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008399.hg.1 |
ENST00000306851.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008399.hg.1 |
ENST00000306851.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008399.hg.1 |
ENST00000306851.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008399.hg.1 |
ENST00000306851.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008399.hg.1 |
ENST00000306851.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008399.hg.1 |
ENST00000306851.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008399.hg.1 |
ENST00000383131.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008399.hg.1 |
ENST00000383131.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008399.hg.1 |
ENST00000383131.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008399.hg.1 |
ENST00000383131.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008399.hg.1 |
ENST00000383131.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008399.hg.1 |
ENST00000383131.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008399.hg.1 |
ENST00000383131.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008399.hg.1 |
ENST00000383131.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008399.hg.1 |
ENST00000383131.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008399.hg.1 |
ENST00000383131.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008399.hg.1 |
ENST00000383131.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008399.hg.1 |
ENST00000578114.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008399.hg.1 |
ENST00000578114.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008399.hg.1 |
ENST00000578114.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008399.hg.1 |
ENST00000578114.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008399.hg.1 |
ENST00000578114.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008399.hg.1 |
ENST00000578114.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008399.hg.1 |
ENST00000578114.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008399.hg.1 |
ENST00000578114.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008399.hg.1 |
ENST00000578114.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008399.hg.1 |
ENST00000578114.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008399.hg.1 |
ENST00000578114.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008399.hg.1 |
ENST00000579372.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008399.hg.1 |
ENST00000579372.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008399.hg.1 |
ENST00000579372.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008399.hg.1 |
NM_004775.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008399.hg.1 |
NM_004775.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008399.hg.1 |
NM_004775.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008399.hg.1 |
NM_004775.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008399.hg.1 |
NM_004775.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008399.hg.1 |
NM_004775.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008399.hg.1 |
NM_004775.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008399.hg.1 |
NM_004775.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008399.hg.1 |
NM_004775.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008399.hg.1 |
NM_004775.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008399.hg.1 |
NM_004775.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008399.hg.1 |
NM_004775.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008399.hg.1 |
NM_004775.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008399.hg.1 |
NM_004775.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008399.hg.1 |
NM_004775.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008399.hg.1 |
NM_004775.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008399.hg.1 |
NM_004775.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004487.hg.1 |
ENST00000237019.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004487.hg.1 |
ENST00000306851.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004487.hg.1 |
ENST00000383131.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004487.hg.1 |
NM_004775.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004487.hg.1 |
TC18000440.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1800157226.hg.1 |
ENST00000237019.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800157226.hg.1 |
ENST00000306851.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800157226.hg.1 |
ENST00000383131.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800157226.hg.1 |
ENST00000578114.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800157226.hg.1 |
NM_004775.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800157226.hg.1 |
TC1800008399.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800157226.hg.1 |
ENST00000237019.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800157226.hg.1 |
ENST00000306851.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800157226.hg.1 |
ENST00000383131.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800157226.hg.1 |
ENST00000578114.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800157226.hg.1 |
NM_004775.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008399.hg.1 |
ENST00000237019.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008399.hg.1 |
ENST00000237019.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008399.hg.1 |
ENST00000237019.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008399.hg.1 |
ENST00000237019.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008399.hg.1 |
ENST00000237019.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008399.hg.1 |
ENST00000237019.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008399.hg.1 |
ENST00000237019.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008399.hg.1 |
ENST00000237019.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008399.hg.1 |
ENST00000237019.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008399.hg.1 |
ENST00000237019.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008399.hg.1 |
ENST00000237019.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008399.hg.1 |
ENST00000237019.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008399.hg.1 |
ENST00000237019.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008399.hg.1 |
ENST00000306851.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008399.hg.1 |
ENST00000306851.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008399.hg.1 |
ENST00000306851.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008399.hg.1 |
ENST00000306851.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008399.hg.1 |
ENST00000306851.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008399.hg.1 |
ENST00000306851.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008399.hg.1 |
ENST00000306851.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008399.hg.1 |
ENST00000306851.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008399.hg.1 |
ENST00000306851.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008399.hg.1 |
ENST00000306851.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008399.hg.1 |
ENST00000306851.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008399.hg.1 |
ENST00000306851.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008399.hg.1 |
ENST00000306851.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008399.hg.1 |
ENST00000306851.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008399.hg.1 |
ENST00000306851.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008399.hg.1 |
ENST00000306851.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008399.hg.1 |
ENST00000383131.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008399.hg.1 |
ENST00000383131.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008399.hg.1 |
ENST00000383131.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008399.hg.1 |
ENST00000383131.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008399.hg.1 |
ENST00000383131.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008399.hg.1 |
ENST00000383131.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008399.hg.1 |
ENST00000383131.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008399.hg.1 |
ENST00000383131.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008399.hg.1 |
ENST00000383131.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008399.hg.1 |
ENST00000383131.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008399.hg.1 |
ENST00000383131.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008399.hg.1 |
ENST00000578114.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008399.hg.1 |
ENST00000578114.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008399.hg.1 |
ENST00000578114.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008399.hg.1 |
ENST00000578114.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008399.hg.1 |
ENST00000578114.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008399.hg.1 |
ENST00000578114.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008399.hg.1 |
ENST00000578114.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008399.hg.1 |
ENST00000578114.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008399.hg.1 |
ENST00000578114.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008399.hg.1 |
ENST00000578114.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008399.hg.1 |
ENST00000578114.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008399.hg.1 |
ENST00000579372.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008399.hg.1 |
ENST00000579372.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008399.hg.1 |
ENST00000579372.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008399.hg.1 |
NM_004775.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008399.hg.1 |
NM_004775.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008399.hg.1 |
NM_004775.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008399.hg.1 |
NM_004775.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008399.hg.1 |
NM_004775.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008399.hg.1 |
NM_004775.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008399.hg.1 |
NM_004775.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008399.hg.1 |
NM_004775.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008399.hg.1 |
NM_004775.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008399.hg.1 |
NM_004775.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008399.hg.1 |
NM_004775.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008399.hg.1 |
NM_004775.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008399.hg.1 |
NM_004775.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008399.hg.1 |
NM_004775.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008399.hg.1 |
NM_004775.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008399.hg.1 |
NM_004775.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008399.hg.1 |
NM_004775.3 |
Refseq |