Assay Details
| Gene Symbol: B4GALNT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173593.3 | NP_775864.3 | 10 - 11 | 991 | 103 | |
| GenBank mRNA | AB089940.1 | - | 10 - 11 | 991 | 103 | |
| AK093503.1 | - | 8 - 9 | 713 | 103 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12000007.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000322843.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000322843.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000322843.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000322843.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000322843.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000322843.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000322843.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000322843.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000322843.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000322843.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000322843.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000322843.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000322843.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000322843.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000535402.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000535402.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000535402.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000535402.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000535402.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000535402.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000535402.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000535402.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000535402.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000535402.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000535402.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000535402.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000535680.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000535680.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000535680.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000535680.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000535680.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000535680.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000535680.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000535680.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000535680.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000540836.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000540836.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000540836.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000540836.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000540836.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000544638.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000544638.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000544638.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000544638.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000544638.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000544638.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000544638.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000544638.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | NM_173593.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | NM_173593.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | NM_173593.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | NM_173593.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | NM_173593.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | NM_173593.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | NM_173593.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | NM_173593.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | NM_173593.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | NM_173593.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | NM_173593.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | NM_173593.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | NM_173593.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | NM_173593.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | NM_173593.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | NM_173593.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | NM_173593.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | NM_173593.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | NM_173593.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | NM_173593.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | NM_173593.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | NM_173593.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | NM_173593.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12000101.hg.1 | ENST00000266383.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12000101.hg.1 | NM_173593.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12000101.hg.1 | TC12000007.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000322843.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000322843.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000322843.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000322843.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000322843.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000322843.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000322843.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000322843.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000322843.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000322843.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000322843.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000322843.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000322843.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000322843.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000535402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000535402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000535402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000535402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000535402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000535402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000535402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000535402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000535402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000535402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000535402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000535402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000535680.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000535680.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000535680.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000535680.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000535680.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000535680.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000535680.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000535680.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000535680.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000540836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000540836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000540836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000540836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000540836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000544638.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000544638.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000544638.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000544638.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000544638.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000544638.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000544638.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000544638.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | NM_173593.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | NM_173593.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | NM_173593.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | NM_173593.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | NM_173593.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | NM_173593.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | NM_173593.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | NM_173593.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | NM_173593.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | NM_173593.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | NM_173593.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | NM_173593.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | NM_173593.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | NM_173593.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | NM_173593.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | NM_173593.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | NM_173593.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | NM_173593.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | NM_173593.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | NM_173593.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | NM_173593.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | NM_173593.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | NM_173593.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200144181.hg.1 | ENST00000266383.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200144181.hg.1 | ENST00000322843.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200144181.hg.1 | NM_173593.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200144181.hg.1 | ENST00000266383.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200144181.hg.1 | ENST00000322843.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200144181.hg.1 | NM_173593.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200144181.hg.1 | TC1200006445.hg.1 | Clariom D TC |

B4GALNT3
Genomic Map