Assay Details
Gene Symbol: B4GALNT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173593.3 | NP_775864.3 | 9 - 10 | 852 | 75 | |
GenBank mRNA | AB089940.1 | - | 9 - 10 | 852 | 75 | |
AK093503.1 | - | 7 - 8 | 574 | 75 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC12000007.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR12000100.hg.1 | ENST00000266383.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12000100.hg.1 | NM_173593.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12000100.hg.1 | TC12000007.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000322843.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000322843.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000322843.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000322843.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000322843.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000322843.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000322843.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000322843.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000322843.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000322843.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000322843.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000322843.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000322843.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000322843.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000535402.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000535402.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000535402.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000535402.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000535402.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000535402.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000535402.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000535402.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000535402.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000535402.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000535402.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000535402.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000535680.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000535680.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000535680.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000535680.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000535680.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000535680.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000535680.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000535680.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000535680.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000540836.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000540836.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000540836.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000540836.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000540836.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000544638.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000544638.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000544638.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000544638.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000544638.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000544638.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000544638.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | ENST00000544638.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | NM_173593.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | NM_173593.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | NM_173593.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | NM_173593.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | NM_173593.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | NM_173593.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | NM_173593.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | NM_173593.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | NM_173593.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | NM_173593.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | NM_173593.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | NM_173593.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | NM_173593.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | NM_173593.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | NM_173593.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | NM_173593.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | NM_173593.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | NM_173593.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | NM_173593.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | NM_173593.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | NM_173593.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | NM_173593.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006445.hg.1 | NM_173593.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12000098.hg.1 | ENST00000266383.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12000098.hg.1 | ENST00000544638.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12000098.hg.1 | NM_173593.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12000098.hg.1 | TC12000007.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1200144180.hg.1 | ENST00000266383.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200144180.hg.1 | ENST00000322843.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200144180.hg.1 | NM_173593.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200144180.hg.1 | ENST00000266383.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200144180.hg.1 | ENST00000322843.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200144180.hg.1 | NM_173593.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200144180.hg.1 | TC1200006445.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000266383.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000322843.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000322843.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000322843.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000322843.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000322843.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000322843.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000322843.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000322843.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000322843.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000322843.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000322843.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000322843.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000322843.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000322843.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000535402.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000535402.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000535402.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000535402.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000535402.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000535402.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000535402.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000535402.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000535402.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000535402.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000535402.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000535402.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000535680.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000535680.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000535680.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000535680.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000535680.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000535680.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000535680.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000535680.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000535680.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000540836.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000540836.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000540836.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000540836.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000540836.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000544638.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000544638.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000544638.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000544638.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000544638.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000544638.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000544638.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | ENST00000544638.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | NM_173593.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | NM_173593.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | NM_173593.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | NM_173593.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | NM_173593.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | NM_173593.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | NM_173593.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | NM_173593.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | NM_173593.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | NM_173593.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | NM_173593.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | NM_173593.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | NM_173593.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | NM_173593.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | NM_173593.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | NM_173593.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | NM_173593.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | NM_173593.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | NM_173593.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | NM_173593.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | NM_173593.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | NM_173593.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006445.hg.1 | NM_173593.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200144178.hg.1 | ENST00000266383.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200144178.hg.1 | ENST00000322843.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200144178.hg.1 | ENST00000544638.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200144178.hg.1 | NM_173593.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200144178.hg.1 | ENST00000266383.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200144178.hg.1 | ENST00000322843.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200144178.hg.1 | ENST00000544638.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200144178.hg.1 | NM_173593.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200144178.hg.1 | TC1200006445.hg.1 | Clariom D TC |
