Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01059428.hg.1 |
TC01003897.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01059428.hg.1 |
ENST00000366812.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01059428.hg.1 |
NM_022735.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01059427.hg.1 |
TC01003897.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01059427.hg.1 |
ENST00000366812.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01059427.hg.1 |
NM_022735.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017519.hg.1 |
ENST00000366812.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017519.hg.1 |
ENST00000366812.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017519.hg.1 |
ENST00000366812.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017519.hg.1 |
ENST00000366812.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017519.hg.1 |
ENST00000366812.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017519.hg.1 |
ENST00000366812.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017519.hg.1 |
ENST00000366812.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017519.hg.1 |
ENST00000366812.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017519.hg.1 |
ENST00000366812.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017519.hg.1 |
ENST00000366812.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017519.hg.1 |
ENST00000366812.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017519.hg.1 |
ENST00000366812.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017519.hg.1 |
ENST00000464927.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017519.hg.1 |
ENST00000464927.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017519.hg.1 |
NM_022735.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017519.hg.1 |
NM_022735.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017519.hg.1 |
NM_022735.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017519.hg.1 |
NM_022735.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017519.hg.1 |
NM_022735.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017519.hg.1 |
NM_022735.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017519.hg.1 |
NM_022735.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017519.hg.1 |
NM_022735.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017519.hg.1 |
NM_022735.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017519.hg.1 |
NM_022735.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017519.hg.1 |
NM_022735.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017519.hg.1 |
NM_022735.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100232592.hg.1 |
ENST00000366812.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100232592.hg.1 |
NM_022735.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100232592.hg.1 |
ENST00000366812.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100232592.hg.1 |
NM_022735.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100232592.hg.1 |
TC0100017519.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100017519.hg.1 |
ENST00000366812.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017519.hg.1 |
ENST00000366812.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017519.hg.1 |
ENST00000366812.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017519.hg.1 |
ENST00000366812.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017519.hg.1 |
ENST00000366812.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017519.hg.1 |
ENST00000366812.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017519.hg.1 |
ENST00000366812.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017519.hg.1 |
ENST00000366812.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017519.hg.1 |
ENST00000366812.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017519.hg.1 |
ENST00000366812.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017519.hg.1 |
ENST00000366812.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017519.hg.1 |
ENST00000366812.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017519.hg.1 |
ENST00000464927.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017519.hg.1 |
ENST00000464927.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017519.hg.1 |
NM_022735.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017519.hg.1 |
NM_022735.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017519.hg.1 |
NM_022735.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017519.hg.1 |
NM_022735.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017519.hg.1 |
NM_022735.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017519.hg.1 |
NM_022735.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017519.hg.1 |
NM_022735.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017519.hg.1 |
NM_022735.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017519.hg.1 |
NM_022735.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017519.hg.1 |
NM_022735.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017519.hg.1 |
NM_022735.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017519.hg.1 |
NM_022735.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100232591.hg.1 |
ENST00000366812.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100232591.hg.1 |
NM_022735.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100232591.hg.1 |
ENST00000366812.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100232591.hg.1 |
NM_022735.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100232591.hg.1 |
TC0100017519.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100232590.hg.1 |
ENST00000366812.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100232590.hg.1 |
NM_022735.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100232590.hg.1 |
ENST00000366812.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100232590.hg.1 |
NM_022735.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100232590.hg.1 |
TC0100017519.hg.1 |
Clariom D TC |