Assay Details
| Gene Symbol: JAG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002226.4 | NP_002217.3 | 19 - 20 | 2795 | 66 | |
| NM_145159.2 | NP_660142.1 | 18 - 19 | 2681 | 66 | ||
| GenBank mRNA | AF003521.1 | - | 19 - 20 | 2423 | 66 | |
| AF020201.1 | - | 19 - 20 | 2442 | 66 | ||
| AF029778.1 | - | 19 - 20 | 2795 | 66 | ||
| AF029779.1 | - | 18 - 19 | 2681 | 66 | ||
| Y14330.1 | - | 19 - 20 | 2346 | 66 | ||
More Information
Set Membership: |
|

JAG2
Genomic Map