Assay Details
Gene Symbol: ZNF790 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001242801.1 | NP_001229730.1 | 1 - 2 | 268 | 113 | |
GenBank mRNA | BF132049.1 | - | 1 - 2 | 165 | 113 | |
DC307556.1 | - | 1 - 2 | 266 | 113 | ||
DC352882.1 | - | 1 - 2 | 314 | 113 | ||
DC377558.1 | - | 1 - 2 | 282 | 113 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC19001472.hg
Additional Information:
This assay is in the 5'UTR region of the NM_001242801.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000356725.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000356725.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000356725.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000356725.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000356725.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000356725.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000356725.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000356725.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000356725.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000356725.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000525288.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000525288.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000525288.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000525288.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000525288.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000527645.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000527645.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000527645.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000528994.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000528994.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000528994.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000528994.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000528994.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000528994.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000528994.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000586323.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000586323.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000586323.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000586323.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000586323.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000613249.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000613249.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000613249.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000613249.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000613249.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000613249.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000613249.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000613249.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000614179.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000614179.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000614179.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000614179.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000614179.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000614179.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000614179.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000614179.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000615484.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000615484.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000615484.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000615484.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000615484.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000615484.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000615484.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000615484.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | NM_001242800.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | NM_001242800.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | NM_001242800.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | NM_001242800.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | NM_001242800.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | NM_001242800.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | NM_001242800.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | NM_001242800.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | NM_001242801.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | NM_001242801.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | NM_001242801.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | NM_001242801.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | NM_001242801.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | NM_001242801.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | NM_001242801.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | NM_001242801.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | NM_001242802.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | NM_001242802.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | NM_001242802.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | NM_001242802.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | NM_001242802.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | NM_001242802.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | NM_001242802.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | NM_001242802.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | NM_206894.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | NM_206894.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | NM_206894.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | NM_206894.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | NM_206894.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | NM_206894.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | NM_206894.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | NM_206894.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19020021.hg.1 | NM_001242801.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19020021.hg.1 | TC19001472.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR19020017.hg.1 | ENST00000356725.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19020017.hg.1 | NM_001242800.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19020017.hg.1 | NM_001242801.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19020017.hg.1 | NM_001242802.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19020017.hg.1 | NM_206894.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19020017.hg.1 | TC19001472.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1900186615.hg.1 | ENST00000528994.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900186615.hg.1 | ENST00000614179.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900186615.hg.1 | NM_001242801.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900186615.hg.1 | TC1900010538.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900186615.hg.1 | ENST00000528994.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900186615.hg.1 | ENST00000614179.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900186615.hg.1 | NM_001242801.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000356725.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000356725.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000356725.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000356725.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000356725.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000356725.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000356725.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000356725.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000356725.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000356725.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000525288.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000525288.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000525288.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000525288.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000525288.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000527645.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000527645.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000527645.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000528994.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000528994.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000528994.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000528994.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000528994.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000528994.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000528994.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000586323.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000586323.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000586323.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000586323.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000586323.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000613249.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000613249.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000613249.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000613249.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000613249.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000613249.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000613249.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000613249.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000614179.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000614179.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000614179.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000614179.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000614179.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000614179.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000614179.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000614179.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000615484.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000615484.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000615484.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000615484.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000615484.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000615484.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000615484.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000615484.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | NM_001242800.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | NM_001242800.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | NM_001242800.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | NM_001242800.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | NM_001242800.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | NM_001242800.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | NM_001242800.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | NM_001242800.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | NM_001242801.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | NM_001242801.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | NM_001242801.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | NM_001242801.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | NM_001242801.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | NM_001242801.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | NM_001242801.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | NM_001242801.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | NM_001242802.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | NM_001242802.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | NM_001242802.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | NM_001242802.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | NM_001242802.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | NM_001242802.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | NM_001242802.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | NM_001242802.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | NM_206894.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | NM_206894.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | NM_206894.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | NM_206894.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | NM_206894.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | NM_206894.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | NM_206894.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | NM_206894.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900186610.hg.1 | ENST00000356725.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900186610.hg.1 | ENST00000525288.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900186610.hg.1 | ENST00000527645.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900186610.hg.1 | ENST00000528994.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900186610.hg.1 | ENST00000586323.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900186610.hg.1 | ENST00000613249.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900186610.hg.1 | ENST00000614179.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900186610.hg.1 | ENST00000615484.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900186610.hg.1 | NM_001242800.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900186610.hg.1 | NM_001242801.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900186610.hg.1 | NM_001242802.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900186610.hg.1 | NM_206894.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900186610.hg.1 | TC1900010538.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900186610.hg.1 | ENST00000356725.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900186610.hg.1 | ENST00000525288.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900186610.hg.1 | ENST00000527645.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900186610.hg.1 | ENST00000528994.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900186610.hg.1 | ENST00000586323.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900186610.hg.1 | ENST00000613249.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900186610.hg.1 | ENST00000614179.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900186610.hg.1 | ENST00000615484.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900186610.hg.1 | NM_001242800.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900186610.hg.1 | NM_001242801.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900186610.hg.1 | NM_001242802.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900186610.hg.1 | NM_206894.3 | Refseq |
