Assay Details
| Gene Symbol: MTSS1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138383.2 | NP_612392.1 | 12 - 13 | 1387 | 66 | |
| XM_005256236.4 | XP_005256293.2 | 11 - 12 | 1438 | 66 | ||
| XM_011523450.2 | XP_011521752.1 | 14 - 15 | 1348 | 66 | ||
| XM_011523453.2 | XP_011521755.2 | 13 - 14 | 1600 | 66 | ||
| XM_011523454.2 | XP_011521756.2 | 12 - 13 | 1591 | 66 | ||
| XM_011523458.2 | XP_011521760.1 | 7 - 8 | 825 | 66 | ||
| XM_017023879.1 | XP_016879368.1 | 13 - 14 | 1504 | 66 | ||
| XM_017023881.1 | XP_016879370.1 | 12 - 13 | 1447 | 66 | ||
| GenBank mRNA | AB115770.1 | - | 12 - 13 | 1127 | 66 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16001237.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR16017076.hg.1 | ENST00000338779.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16017076.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16017076.hg.1 | TC16001237.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000562883.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000562883.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000562883.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000562883.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000562883.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000576338.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000576338.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000576338.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000576338.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000576338.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000576338.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000576338.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000616026.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000616026.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000616026.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000616026.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000616026.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000616026.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000616026.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000616026.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000616026.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000616026.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000616026.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000616026.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000616026.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000616026.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000616026.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000616026.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000616026.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000616026.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16017077.hg.1 | ENST00000338779.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16017077.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16017077.hg.1 | TC16001237.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000562883.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000562883.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000562883.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000562883.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000562883.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000576338.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000576338.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000576338.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000576338.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000576338.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000576338.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000576338.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000616026.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000616026.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000616026.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000616026.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000616026.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000616026.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000616026.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000616026.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000616026.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000616026.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000616026.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000616026.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000616026.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000616026.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000616026.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000616026.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000616026.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000616026.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600194266.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600194266.hg.1 | ENST00000616026.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600194266.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600194266.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600194266.hg.1 | ENST00000616026.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600194266.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600194266.hg.1 | TC1600011560.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600194265.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600194265.hg.1 | ENST00000616026.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600194265.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600194265.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600194265.hg.1 | ENST00000616026.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600194265.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600194265.hg.1 | TC1600011560.hg.1 | Clariom D TC |

MTSS1L
Genomic Map