Assay Details
| Gene Symbol: CRLF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015986.3 | NP_057070.3 | 1 - 2 | 230 | 69 | |
| GenBank mRNA | AF120151.1 | - | 596 | 69 | ||
| AK313315.1 | - | 1 - 2 | 219 | 69 | ||
| BC023567.2 | - | 1 - 2 | 152 | 4309046 | 69 | |
| BU902521.1 | - | 1 - 2 | 271 | 69 | ||
| DQ298450.1 | - | 1 - 2 | 109 | 69 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17001336.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
G1/S transition of mitotic cell cycle
negative regulation of cell growth
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of JAK-STAT cascade
positive regulation of cell cycle arrest
protein binding
G1/S transition of mitotic cell cycle
negative regulation of cell growth
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of JAK-STAT cascade
positive regulation of cell cycle arrest
protein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR17017811.hg.1 | ENST00000324238.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17017811.hg.1 | NM_015986.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17017811.hg.1 | TC17001336.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011565.hg.1 | ENST00000511386.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | ENST00000324238.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | ENST00000324238.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | ENST00000324238.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | ENST00000324238.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | ENST00000324238.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | ENST00000324238.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | ENST00000324238.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | ENST00000324238.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | ENST00000324238.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | ENST00000324238.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | ENST00000324238.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | ENST00000324238.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | ENST00000324238.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | ENST00000324238.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | ENST00000324238.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | ENST00000578692.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | ENST00000578692.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | ENST00000578692.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | ENST00000578692.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | ENST00000578692.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | ENST00000578692.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | ENST00000578692.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | ENST00000578692.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | ENST00000578692.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | ENST00000578692.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | ENST00000578692.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | ENST00000583527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | ENST00000583527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | ENST00000583527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | ENST00000583527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | ENST00000583527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | ENST00000583805.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | ENST00000583805.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | ENST00000585268.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | ENST00000585268.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | ENST00000585268.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | ENST00000585268.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | NM_015986.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | NM_015986.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | NM_015986.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | NM_015986.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | NM_015986.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | NM_015986.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | NM_015986.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | NM_015986.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | NM_015986.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | NM_015986.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | NM_015986.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | NM_015986.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | NM_015986.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | NM_015986.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | NM_015986.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | NR_073118.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | NR_073118.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | NR_073118.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | NR_073118.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | NR_073118.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | NR_073118.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | NR_073118.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | NR_073118.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | NR_073118.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | NR_073118.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | NR_073118.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | NR_073118.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | NR_073118.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17017812.hg.1 | ENST00000324238.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17017812.hg.1 | ENST00000544695.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17017812.hg.1 | NM_015986.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17017812.hg.1 | TC17001336.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17017814.hg.1 | ENST00000324238.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17017814.hg.1 | NM_015986.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17017814.hg.1 | TC17001336.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008695.hg.1 | ENST00000440950.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008695.hg.1 | ENST00000440950.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008695.hg.1 | ENST00000440950.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008695.hg.1 | ENST00000440950.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011565.hg.1 | ENST00000511386.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700180959.hg.1 | ENST00000324238.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700180959.hg.1 | ENST00000583805.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700180959.hg.1 | NM_015986.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700180959.hg.1 | ENST00000324238.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700180959.hg.1 | ENST00000583805.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700180959.hg.1 | NM_015986.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700180959.hg.1 | TC1700010293.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700180961.hg.1 | ENST00000324238.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700180961.hg.1 | ENST00000578692.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700180961.hg.1 | NM_015986.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700180961.hg.1 | NR_073118.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700180961.hg.1 | ENST00000324238.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700180961.hg.1 | ENST00000578692.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700180961.hg.1 | NM_015986.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700180961.hg.1 | NR_073118.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700180961.hg.1 | TC1700010293.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | ENST00000324238.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | ENST00000324238.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | ENST00000324238.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | ENST00000324238.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | ENST00000324238.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | ENST00000324238.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | ENST00000324238.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | ENST00000324238.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | ENST00000324238.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | ENST00000324238.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | ENST00000324238.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | ENST00000324238.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | ENST00000324238.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | ENST00000324238.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | ENST00000324238.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | ENST00000578692.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | ENST00000578692.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | ENST00000578692.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | ENST00000578692.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | ENST00000578692.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | ENST00000578692.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | ENST00000578692.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | ENST00000578692.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | ENST00000578692.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | ENST00000578692.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | ENST00000578692.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | ENST00000583527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | ENST00000583527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | ENST00000583527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | ENST00000583527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | ENST00000583527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | ENST00000583805.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | ENST00000583805.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | ENST00000585268.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | ENST00000585268.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | ENST00000585268.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | ENST00000585268.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | NM_015986.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | NM_015986.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | NM_015986.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | NM_015986.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | NM_015986.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | NM_015986.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | NM_015986.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | NM_015986.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | NM_015986.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | NM_015986.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | NM_015986.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | NM_015986.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | NM_015986.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | NM_015986.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | NM_015986.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | NR_073118.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | NR_073118.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | NR_073118.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | NR_073118.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | NR_073118.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | NR_073118.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | NR_073118.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | NR_073118.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | NR_073118.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | NR_073118.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | NR_073118.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | NR_073118.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | NR_073118.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700180964.hg.1 | ENST00000324238.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700180964.hg.1 | NM_015986.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700180964.hg.1 | NR_073118.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700180964.hg.1 | ENST00000324238.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700180964.hg.1 | NM_015986.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700180964.hg.1 | NR_073118.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700180964.hg.1 | TC1700010293.hg.1 | Clariom D TC |

CRLF3
Genomic Map