Assay Details
Gene Symbol: OPRPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001302807.1 | NP_001289736.1 | 1 - 2 | 165 | 118 | |
NM_021225.4 | NP_067048.4 | 1 - 2 | 165 | 118 | ||
GenBank mRNA | CD722615.1 | - | 1 - 2 | 165 | 118 | |
CD723077.1 | - | 1 - 2 | 92 | 118 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC04000385.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_001302807.1,NM_021225.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR04005241.hg.1 | TC04000385.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04005241.hg.1 | ENST00000399575.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04005241.hg.1 | NM_021225.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04005243.hg.1 | TC04000385.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04005243.hg.1 | ENST00000399575.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04005243.hg.1 | ENST00000514338.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04005243.hg.1 | NM_021225.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007781.hg.1 | ENST00000399575.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007781.hg.1 | ENST00000399575.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007781.hg.1 | ENST00000399575.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007781.hg.1 | ENST00000399575.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007781.hg.1 | ENST00000399575.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007781.hg.1 | ENST00000399575.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007781.hg.1 | ENST00000399575.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007781.hg.1 | ENST00000505023.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007781.hg.1 | ENST00000505023.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007781.hg.1 | ENST00000514338.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007781.hg.1 | ENST00000514338.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007781.hg.1 | ENST00000514338.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007781.hg.1 | ENST00000514338.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007781.hg.1 | ENST00000514338.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007781.hg.1 | ENST00000615728.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007781.hg.1 | ENST00000615728.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007781.hg.1 | ENST00000615728.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007781.hg.1 | ENST00000615728.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007781.hg.1 | ENST00000615728.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007781.hg.1 | NM_001302807.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007781.hg.1 | NM_001302807.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007781.hg.1 | NM_001302807.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007781.hg.1 | NM_001302807.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007781.hg.1 | NM_001302807.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007781.hg.1 | NM_001302807.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007781.hg.1 | NM_001302807.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007781.hg.1 | NM_001302807.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007781.hg.1 | NM_021225.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007781.hg.1 | NM_021225.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007781.hg.1 | NM_021225.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007781.hg.1 | NM_021225.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007781.hg.1 | NM_021225.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007781.hg.1 | NM_021225.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007781.hg.1 | NM_021225.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007781.hg.1 | NR_126503.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007781.hg.1 | NR_126503.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007781.hg.1 | NR_126503.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007781.hg.1 | NR_126503.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007781.hg.1 | NR_126503.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007781.hg.1 | NR_126503.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04005242.hg.1 | TC04000385.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04005242.hg.1 | ENST00000399575.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04005242.hg.1 | ENST00000514338.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04005242.hg.1 | NM_021225.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04005245.hg.1 | TC04000385.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04005245.hg.1 | ENST00000399575.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04005245.hg.1 | ENST00000514338.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04005245.hg.1 | NM_021225.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400153118.hg.1 | ENST00000514338.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400153118.hg.1 | NM_001302807.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400153118.hg.1 | ENST00000514338.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400153118.hg.1 | NM_001302807.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400153118.hg.1 | TC0400007781.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0400153112.hg.1 | ENST00000399575.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400153112.hg.1 | NM_001302807.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400153112.hg.1 | NM_021225.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400153112.hg.1 | NR_126503.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400153112.hg.1 | ENST00000399575.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400153112.hg.1 | NM_001302807.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400153112.hg.1 | NM_021225.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400153112.hg.1 | NR_126503.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400153112.hg.1 | TC0400007781.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0400153117.hg.1 | ENST00000399575.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400153117.hg.1 | ENST00000514338.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400153117.hg.1 | ENST00000615728.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400153117.hg.1 | NM_001302807.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400153117.hg.1 | NM_021225.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400153117.hg.1 | ENST00000399575.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400153117.hg.1 | ENST00000514338.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400153117.hg.1 | ENST00000615728.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400153117.hg.1 | NM_001302807.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400153117.hg.1 | NM_021225.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400153117.hg.1 | TC0400007781.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0400153113.hg.1 | ENST00000399575.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400153113.hg.1 | ENST00000514338.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400153113.hg.1 | NM_001302807.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400153113.hg.1 | NM_021225.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400153113.hg.1 | NR_126503.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400153113.hg.1 | ENST00000399575.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400153113.hg.1 | ENST00000514338.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400153113.hg.1 | NM_001302807.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400153113.hg.1 | NM_021225.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400153113.hg.1 | NR_126503.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400153113.hg.1 | TC0400007781.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0400153115.hg.1 | ENST00000399575.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400153115.hg.1 | ENST00000505023.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400153115.hg.1 | ENST00000514338.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400153115.hg.1 | ENST00000615728.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400153115.hg.1 | NM_001302807.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400153115.hg.1 | NM_021225.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400153115.hg.1 | NR_126503.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400153115.hg.1 | ENST00000399575.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400153115.hg.1 | ENST00000505023.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400153115.hg.1 | ENST00000514338.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400153115.hg.1 | ENST00000615728.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400153115.hg.1 | NM_001302807.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400153115.hg.1 | NM_021225.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400153115.hg.1 | NR_126503.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400153115.hg.1 | TC0400007781.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0400007781.hg.1 | ENST00000399575.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007781.hg.1 | ENST00000399575.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007781.hg.1 | ENST00000399575.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007781.hg.1 | ENST00000399575.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007781.hg.1 | ENST00000399575.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007781.hg.1 | ENST00000399575.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007781.hg.1 | ENST00000399575.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007781.hg.1 | ENST00000505023.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007781.hg.1 | ENST00000505023.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007781.hg.1 | ENST00000514338.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007781.hg.1 | ENST00000514338.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007781.hg.1 | ENST00000514338.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007781.hg.1 | ENST00000514338.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007781.hg.1 | ENST00000514338.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007781.hg.1 | ENST00000615728.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007781.hg.1 | ENST00000615728.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007781.hg.1 | ENST00000615728.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007781.hg.1 | ENST00000615728.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007781.hg.1 | ENST00000615728.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007781.hg.1 | NM_001302807.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007781.hg.1 | NM_001302807.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007781.hg.1 | NM_001302807.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007781.hg.1 | NM_001302807.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007781.hg.1 | NM_001302807.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007781.hg.1 | NM_001302807.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007781.hg.1 | NM_001302807.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007781.hg.1 | NM_001302807.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007781.hg.1 | NM_021225.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007781.hg.1 | NM_021225.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007781.hg.1 | NM_021225.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007781.hg.1 | NM_021225.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007781.hg.1 | NM_021225.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007781.hg.1 | NM_021225.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007781.hg.1 | NM_021225.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007781.hg.1 | NR_126503.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007781.hg.1 | NR_126503.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007781.hg.1 | NR_126503.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007781.hg.1 | NR_126503.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007781.hg.1 | NR_126503.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007781.hg.1 | NR_126503.1 | Refseq |
