Assay Details
| Gene Symbol: STAB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017564.9 | NP_060034.9 | 19 - 20 | 2275 | 77 | |
| XM_011538537.2 | XP_011536839.1 | 19 - 20 | 2316 | 77 | ||
| XM_011538538.2 | XP_011536840.1 | 19 - 20 | 2316 | 77 | ||
| XM_011538539.2 | XP_011536841.1 | 5 - 6 | 519 | 77 | ||
| XM_017019585.1 | XP_016875074.1 | 19 - 20 | 2316 | 77 | ||
| GenBank mRNA | AB052958.1 | - | 19 - 20 | 2271 | 77 | |
| AJ295695.1 | - | 19 - 20 | 2275 | 77 | ||
| AK160380.1 | - | 18 - 19 | 1988 | 77 | ||
| AY311388.1 | - | 19 - 20 | 2293 | 77 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12000799.hg.1
Set Membership: |
|

STAB2
Genomic Map