Assay Details
| Gene Symbol: STAB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017564.9 | NP_060034.9 | 20 - 21 | 2353 | 57 | |
| XM_011538537.2 | XP_011536839.1 | 20 - 21 | 2394 | 57 | ||
| XM_011538538.2 | XP_011536840.1 | 20 - 21 | 2394 | 57 | ||
| XM_011538539.2 | XP_011536841.1 | 6 - 7 | 597 | 57 | ||
| XM_017019585.1 | XP_016875074.1 | 20 - 21 | 2394 | 57 | ||
| GenBank mRNA | AB052958.1 | - | 20 - 21 | 2349 | 57 | |
| AJ295695.1 | - | 20 - 21 | 2353 | 57 | ||
| AK160380.1 | - | 19 - 20 | 2066 | 57 | ||
| AY311388.1 | - | 20 - 21 | 2371 | 57 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12000799.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
angiogenesis
endocytosis
receptor-mediated endocytosis
cell adhesion
regulation of gene expression
regulation of blood coagulation
hyaluronan catabolic process
defense response to bacterium
oxidation-reduction process
low-density lipoprotein receptor activity
scavenger receptor activity
calcium ion binding
protein binding
hyaluronic acid binding
protein disulfide oxidoreductase activity
low-density lipoprotein particle binding
angiogenesis
endocytosis
receptor-mediated endocytosis
cell adhesion
regulation of gene expression
regulation of blood coagulation
hyaluronan catabolic process
defense response to bacterium
oxidation-reduction process
low-density lipoprotein receptor activity
scavenger receptor activity
calcium ion binding
protein binding
hyaluronic acid binding
protein disulfide oxidoreductase activity
low-density lipoprotein particle binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000388887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000388887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000388887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000388887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000388887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000388887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000388887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000388887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000388887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000388887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000388887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000388887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000388887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000388887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000388887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000388887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000388887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000388887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000388887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000388887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000388887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000388887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000388887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000388887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000388887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000388887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000388887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000388887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000388887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000388887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000388887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000388887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000388887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000388887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000388887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000388887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000388887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000388887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000388887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000388887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000388887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000388887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000388887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000388887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000388887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000388887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000388887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000388887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000388887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000388887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000388887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000388887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000388887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000388887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000388887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000388887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000388887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000388887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000388887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000388887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000388887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000388887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000388887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000388887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000388887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000388887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000388887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000388887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000388887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000388887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000388887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000388887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000388887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000388887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000388887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000548073.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000548073.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000548073.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000548073.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000548073.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000548579.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000548579.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000548579.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000548579.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000548579.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000549474.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000549474.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000549474.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000549474.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000549798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000549798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000549798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000552777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000552777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000552777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | ENST00000552777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | NM_017564.9 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | NM_017564.9 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | NM_017564.9 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | NM_017564.9 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | NM_017564.9 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | NM_017564.9 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | NM_017564.9 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | NM_017564.9 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | NM_017564.9 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | NM_017564.9 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | NM_017564.9 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | NM_017564.9 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | NM_017564.9 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | NM_017564.9 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | NM_017564.9 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | NM_017564.9 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | NM_017564.9 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | NM_017564.9 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | NM_017564.9 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | NM_017564.9 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | NM_017564.9 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | NM_017564.9 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | NM_017564.9 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | NM_017564.9 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | NM_017564.9 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | NM_017564.9 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | NM_017564.9 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | NM_017564.9 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | NM_017564.9 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | NM_017564.9 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | NM_017564.9 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | NM_017564.9 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | NM_017564.9 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | NM_017564.9 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | NM_017564.9 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | NM_017564.9 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | NM_017564.9 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | NM_017564.9 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | NM_017564.9 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | NM_017564.9 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | NM_017564.9 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | NM_017564.9 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | NM_017564.9 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | NM_017564.9 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | NM_017564.9 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | NM_017564.9 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | NM_017564.9 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | NM_017564.9 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | NM_017564.9 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | NM_017564.9 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | NM_017564.9 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | NM_017564.9 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | NM_017564.9 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | NM_017564.9 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | NM_017564.9 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | NM_017564.9 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | NM_017564.9 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | NM_017564.9 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | NM_017564.9 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | NM_017564.9 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | NM_017564.9 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | NM_017564.9 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | NM_017564.9 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | NM_017564.9 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | NM_017564.9 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | NM_017564.9 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | NM_017564.9 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | NM_017564.9 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | NM_017564.9 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | NM_017564.9 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | NM_017564.9 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | NM_017564.9 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | NM_017564.9 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | NM_017564.9 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008659.hg.1 | NM_017564.9 | Refseq |

STAB2
Genomic Map