Assay Details
| Gene Symbol: STAB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017564.9 | NP_060034.9 | 39 - 40 | 4479 | 56 | |
| XM_011538537.2 | XP_011536839.1 | 39 - 40 | 4520 | 56 | ||
| XM_011538538.2 | XP_011536840.1 | 39 - 40 | 4520 | 56 | ||
| XM_011538539.2 | XP_011536841.1 | 25 - 26 | 2723 | 56 | ||
| XM_011538541.2 | XP_011536843.1 | 17 - 18 | 1830 | 56 | ||
| XM_011538542.2 | XP_011536844.1 | 6 - 7 | 639 | 56 | ||
| XM_017019585.1 | XP_016875074.1 | 39 - 40 | 4520 | 56 | ||
| GenBank mRNA | AB052958.1 | - | 39 - 40 | 4475 | 56 | |
| AJ295695.1 | - | 39 - 40 | 4479 | 56 | ||
| AK024503.1 | - | 7 - 8 | 821 | 56 | ||
| AK074051.1 | - | 17 - 18 | 1850 | 56 | ||
| AK160380.1 | - | 38 - 39 | 4192 | 56 | ||
| AY227444.1 | - | 8 - 9 | 888 | 56 | ||
| AY311388.1 | - | 39 - 40 | 4497 | 56 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12000799.hg.1
Set Membership: |
|

STAB2
Genomic Map