Assay Details
| Gene Symbol: STAB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017564.9 | NP_060034.9 | 50 - 51 | 5473 | 107 | |
| XM_011538537.2 | XP_011536839.1 | 50 - 51 | 5514 | 107 | ||
| XM_011538538.2 | XP_011536840.1 | 50 - 51 | 5514 | 107 | ||
| XM_011538539.2 | XP_011536841.1 | 36 - 37 | 3717 | 107 | ||
| XM_011538541.2 | XP_011536843.1 | 28 - 29 | 2824 | 107 | ||
| XM_011538542.2 | XP_011536844.1 | 17 - 18 | 1633 | 107 | ||
| XM_017019585.1 | XP_016875074.1 | 50 - 51 | 5514 | 107 | ||
| GenBank mRNA | AB052958.1 | - | 50 - 51 | 5469 | 107 | |
| AF160476.1 | - | 5 - 6 | 495 | 107 | ||
| AJ295695.1 | - | 50 - 51 | 5473 | 107 | ||
| AK024503.1 | - | 18 - 19 | 1815 | 107 | ||
| AK074051.1 | - | 28 - 29 | 2844 | 107 | ||
| AK160380.1 | - | 49 - 50 | 5191 | 107 | ||
| AL133021.1 | - | 10 - 11 | 955 | 107 | ||
| AY227444.1 | - | 19 - 20 | 1882 | 107 | ||
| AY311388.1 | - | 50 - 51 | 5491 | 107 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12000799.hg.1
Set Membership: |
|

STAB2
Genomic Map