Assay Details
| Gene Symbol: STAB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017564.9 | NP_060034.9 | 58 - 59 | 6431 | 65 | |
| XM_011538537.2 | XP_011536839.1 | 58 - 59 | 6472 | 65 | ||
| XM_011538538.2 | XP_011536840.1 | 58 - 59 | 6472 | 65 | ||
| XM_011538539.2 | XP_011536841.1 | 44 - 45 | 4675 | 65 | ||
| XM_011538541.2 | XP_011536843.1 | 36 - 37 | 3782 | 65 | ||
| XM_011538542.2 | XP_011536844.1 | 25 - 26 | 2591 | 65 | ||
| XM_017019585.1 | XP_016875074.1 | 58 - 59 | 6472 | 65 | ||
| GenBank mRNA | AB052958.1 | - | 58 - 59 | 6427 | 65 | |
| AF160476.1 | - | 13 - 14 | 1453 | 65 | ||
| AJ295695.1 | - | 58 - 59 | 6431 | 65 | ||
| AK024503.1 | - | 26 - 27 | 2773 | 65 | ||
| AK074051.1 | - | 36 - 37 | 3802 | 65 | ||
| AK160380.1 | - | 57 - 58 | 6149 | 65 | ||
| AL133021.1 | - | 18 - 19 | 1913 | 65 | ||
| AY227444.1 | - | 27 - 28 | 2840 | 65 | ||
| AY311388.1 | - | 58 - 59 | 6449 | 65 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12000799.hg.1
Set Membership: |
|

STAB2
Genomic Map