Assay Details
| Gene Symbol: STAB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017564.9 | NP_060034.9 | 62 - 63 | 7063 | 76 | |
| XM_011538539.2 | XP_011536841.1 | 48 - 49 | 5307 | 76 | ||
| XM_011538541.2 | XP_011536843.1 | 40 - 41 | 4414 | 76 | ||
| XM_011538542.2 | XP_011536844.1 | 29 - 30 | 3223 | 76 | ||
| GenBank mRNA | AB052958.1 | - | 62 - 63 | 7059 | 76 | |
| AF160476.1 | - | 17 - 18 | 2085 | 76 | ||
| AJ295695.1 | - | 62 - 63 | 7063 | 76 | ||
| AK024503.1 | - | 30 - 31 | 3405 | 76 | ||
| AK074051.1 | - | 40 - 41 | 4434 | 76 | ||
| AK160380.1 | - | 61 - 62 | 6781 | 76 | ||
| AY227444.1 | - | 31 - 32 | 3472 | 76 | ||
| AY311388.1 | - | 62 - 63 | 7081 | 76 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12000799.hg.1
Set Membership: |
|

STAB2
Genomic Map