Assay Details
| Gene Symbol: STAB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017564.9 | NP_060034.9 | 66 - 67 | 7551 | 67 | |
| XM_011538539.2 | XP_011536841.1 | 52 - 53 | 5795 | 67 | ||
| XM_011538541.2 | XP_011536843.1 | 44 - 45 | 4902 | 67 | ||
| XM_011538542.2 | XP_011536844.1 | 33 - 34 | 3711 | 67 | ||
| GenBank mRNA | AB052958.1 | - | 66 - 67 | 7547 | 67 | |
| AF160476.1 | - | 21 - 22 | 2573 | 67 | ||
| AJ295695.1 | - | 66 - 67 | 7551 | 67 | ||
| AK024503.1 | - | 34 - 35 | 3893 | 67 | ||
| AK074051.1 | - | 44 - 45 | 4922 | 67 | ||
| AK160380.1 | - | 65 - 66 | 7269 | 67 | ||
| AL133021.1 | - | 25 - 26 | 2926 | 67 | ||
| AY227444.1 | - | 35 - 36 | 3960 | 67 | ||
| AY311388.1 | - | 66 - 67 | 7569 | 67 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12000799.hg.1
Set Membership: |
|

STAB2
Genomic Map