Assay Details
| Gene Symbol: FUT10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AJ431184.1 | - | 1 - 2 | 250 | 122 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AJ431184.1 transcripts.
Set Membership: |
|

FUT10
Genomic Map