Assay Details
Gene Symbol: NAGPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016256.3 | NP_057340.2 | 2 - 3 | 567 | 152 | |
XM_011522517.2 | XP_011520819.1 | 2 - 3 | 567 | 152 | ||
GenBank mRNA | AF187072.1 | - | 2 - 3 | 540 | 152 | |
AK025570.1 | - | 2 - 3 | 566 | 152 | ||
AK026122.1 | - | 2 - 3 | 543 | 152 | ||
AK127952.1 | - | 2 - 3 | 561 | 152 | ||
AK314320.1 | - | 2 - 3 | 561 | 152 | ||
BC012194.1 | - | 2 - 3 | 557 | 4661335 | 152 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC16000840.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000312251.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000312251.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000312251.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000312251.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000312251.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000312251.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000312251.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000312251.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000312251.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000312251.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000312251.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000312251.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000312251.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000312251.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000312251.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000312251.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000312251.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000312251.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000312251.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000312251.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000312251.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000381955.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000381955.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000381955.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000381955.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000381955.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000381955.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000381955.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000381955.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000381955.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000381955.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000381955.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000381955.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000381955.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000381955.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000381955.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000381955.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000381955.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000381955.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000381955.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000381955.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000561580.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000561580.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000561580.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000561580.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000561580.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000561580.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000562037.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000562037.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000562037.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000562037.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000562037.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000562037.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000562346.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000562346.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000562346.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000562346.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000562346.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000562746.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000562746.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000562746.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000562746.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000562746.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000562746.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000562746.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000562746.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000562746.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000562746.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000562746.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000562746.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000562746.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000562746.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000562746.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000562746.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000562746.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000562746.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000562746.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000562746.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000562746.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000562746.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000563578.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000563578.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000563578.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000563578.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000563578.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000563578.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000563578.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000563578.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000563578.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000563578.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000563578.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000564397.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000564397.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000564397.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000564397.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000564397.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000564397.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000564397.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000564397.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000564397.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000564397.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000564397.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000564397.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000564397.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000564397.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000564397.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000564397.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000564397.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000564397.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000564397.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000564397.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000564922.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000564922.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000564922.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000564922.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000565876.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000565876.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000565876.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000565876.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000565876.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000565876.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000565876.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000566137.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000566137.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000566137.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000566137.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000566137.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000566137.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000566137.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000566137.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000567739.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000567739.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000567739.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000567739.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000567739.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000567739.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000567739.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000567739.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000568202.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000568202.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000568202.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000568202.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000568202.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000568202.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000568202.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000568202.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000568202.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000568202.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000568202.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000568202.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000568202.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000568202.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000568202.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000568528.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000568528.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000568528.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000568528.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000568528.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000568528.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000569296.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000569296.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000569296.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000569296.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000569296.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000569296.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000569296.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000569296.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000569296.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000569296.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000569296.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000569296.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000569296.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000569793.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000569793.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000569793.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000569793.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000591478.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000591478.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | ENST00000591478.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | NM_016256.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | NM_016256.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | NM_016256.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | NM_016256.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | NM_016256.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | NM_016256.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | NM_016256.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | NM_016256.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | NM_016256.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | NM_016256.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | NM_016256.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | NM_016256.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | NM_016256.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | NM_016256.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | NM_016256.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | NM_016256.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | NM_016256.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | NM_016256.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | NM_016256.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | NM_016256.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009256.hg.1 | NM_016256.3 | Refseq |
