Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19008560.hg.1 |
ENST00000426739.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19008560.hg.1 |
NM_001144824.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19008560.hg.1 |
NM_006630.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19008560.hg.1 |
TC19000619.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19008559.hg.1 |
ENST00000426739.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19008559.hg.1 |
NM_006630.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19008559.hg.1 |
TC19000619.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008254.hg.1 |
ENST00000426739.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008254.hg.1 |
ENST00000426739.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008254.hg.1 |
ENST00000426739.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008254.hg.1 |
ENST00000426739.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008254.hg.1 |
ENST00000426739.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008254.hg.1 |
ENST00000426739.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008254.hg.1 |
ENST00000426739.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008254.hg.1 |
ENST00000426739.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008254.hg.1 |
ENST00000426739.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008254.hg.1 |
ENST00000590748.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008254.hg.1 |
ENST00000590748.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008254.hg.1 |
ENST00000590748.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008254.hg.1 |
ENST00000590795.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008254.hg.1 |
ENST00000590795.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008254.hg.1 |
ENST00000590795.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008254.hg.1 |
ENST00000590795.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008254.hg.1 |
ENST00000590795.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008254.hg.1 |
ENST00000590795.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008254.hg.1 |
ENST00000590795.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008254.hg.1 |
ENST00000592437.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008254.hg.1 |
ENST00000592437.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008254.hg.1 |
ENST00000592437.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008254.hg.1 |
ENST00000592437.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008254.hg.1 |
ENST00000592437.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008254.hg.1 |
ENST00000592437.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008254.hg.1 |
ENST00000592437.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008254.hg.1 |
ENST00000592437.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008254.hg.1 |
NM_001144824.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008254.hg.1 |
NM_001144824.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008254.hg.1 |
NM_001144824.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008254.hg.1 |
NM_001144824.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008254.hg.1 |
NM_001144824.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008254.hg.1 |
NM_001144824.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008254.hg.1 |
NM_001144824.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008254.hg.1 |
NM_001144824.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008254.hg.1 |
NM_001144824.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008254.hg.1 |
NM_006630.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008254.hg.1 |
NM_006630.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008254.hg.1 |
NM_006630.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008254.hg.1 |
NM_006630.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008254.hg.1 |
NM_006630.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008254.hg.1 |
NM_006630.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008254.hg.1 |
NM_006630.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008254.hg.1 |
NM_006630.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008254.hg.1 |
NM_006630.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008254.hg.1 |
NM_006630.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008254.hg.1 |
ENST00000426739.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008254.hg.1 |
ENST00000426739.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008254.hg.1 |
ENST00000426739.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008254.hg.1 |
ENST00000426739.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008254.hg.1 |
ENST00000426739.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008254.hg.1 |
ENST00000426739.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008254.hg.1 |
ENST00000426739.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008254.hg.1 |
ENST00000426739.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008254.hg.1 |
ENST00000426739.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008254.hg.1 |
ENST00000590748.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008254.hg.1 |
ENST00000590748.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008254.hg.1 |
ENST00000590748.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008254.hg.1 |
ENST00000590795.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008254.hg.1 |
ENST00000590795.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008254.hg.1 |
ENST00000590795.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008254.hg.1 |
ENST00000590795.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008254.hg.1 |
ENST00000590795.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008254.hg.1 |
ENST00000590795.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008254.hg.1 |
ENST00000590795.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008254.hg.1 |
ENST00000592437.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008254.hg.1 |
ENST00000592437.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008254.hg.1 |
ENST00000592437.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008254.hg.1 |
ENST00000592437.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008254.hg.1 |
ENST00000592437.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008254.hg.1 |
ENST00000592437.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008254.hg.1 |
ENST00000592437.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008254.hg.1 |
ENST00000592437.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008254.hg.1 |
NM_001144824.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008254.hg.1 |
NM_001144824.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008254.hg.1 |
NM_001144824.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008254.hg.1 |
NM_001144824.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008254.hg.1 |
NM_001144824.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008254.hg.1 |
NM_001144824.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008254.hg.1 |
NM_001144824.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008254.hg.1 |
NM_001144824.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008254.hg.1 |
NM_001144824.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008254.hg.1 |
NM_006630.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008254.hg.1 |
NM_006630.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008254.hg.1 |
NM_006630.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008254.hg.1 |
NM_006630.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008254.hg.1 |
NM_006630.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008254.hg.1 |
NM_006630.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008254.hg.1 |
NM_006630.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008254.hg.1 |
NM_006630.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008254.hg.1 |
NM_006630.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008254.hg.1 |
NM_006630.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900163007.hg.1 |
ENST00000426739.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900163007.hg.1 |
ENST00000590795.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900163007.hg.1 |
NM_006630.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900163007.hg.1 |
TC1900008254.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900163007.hg.1 |
ENST00000426739.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900163007.hg.1 |
ENST00000590795.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900163007.hg.1 |
NM_006630.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900163009.hg.1 |
ENST00000426739.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900163009.hg.1 |
ENST00000590748.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900163009.hg.1 |
ENST00000590795.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900163009.hg.1 |
ENST00000592437.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900163009.hg.1 |
NM_001144824.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900163009.hg.1 |
NM_006630.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900163009.hg.1 |
TC1900008254.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900163009.hg.1 |
ENST00000426739.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900163009.hg.1 |
ENST00000590748.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900163009.hg.1 |
ENST00000590795.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900163009.hg.1 |
ENST00000592437.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900163009.hg.1 |
NM_001144824.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900163009.hg.1 |
NM_006630.2 |
Refseq |