Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
ENST00000373547.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
ENST00000373547.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
ENST00000373547.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
ENST00000373547.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
ENST00000373547.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
ENST00000373547.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
ENST00000373547.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
ENST00000373547.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
ENST00000373547.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
ENST00000373547.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
ENST00000373547.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
ENST00000373547.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
ENST00000415905.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
ENST00000415905.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
ENST00000415905.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
ENST00000415905.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
ENST00000415905.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
ENST00000415905.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
ENST00000415905.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
ENST00000415905.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
ENST00000415905.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
ENST00000415905.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
ENST00000415905.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
ENST00000415905.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
ENST00000451402.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
ENST00000451402.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
ENST00000451402.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
ENST00000451402.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
ENST00000451402.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
ENST00000451402.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
ENST00000451402.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
ENST00000451402.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
ENST00000451402.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
ENST00000451402.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
ENST00000451402.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
ENST00000451402.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
ENST00000451402.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
ENST00000451402.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
ENST00000451402.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
ENST00000456642.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
ENST00000456642.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
ENST00000456642.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
ENST00000456642.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
ENST00000456642.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
ENST00000456642.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
ENST00000456642.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
NM_001123355.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
NM_001123355.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
NM_001123355.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
NM_001123355.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
NM_001123355.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
NM_001123355.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
NM_001123355.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
NM_001123355.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
NM_001123355.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
NM_001123355.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
NM_001123355.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
NM_001123355.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
NM_001123355.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
NM_001123355.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
NM_001123355.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
NM_001123369.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
NM_001123369.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
NM_001123369.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
NM_001123369.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
NM_001123369.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
NM_001123369.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
NM_001123369.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
NM_001123369.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
NM_001123369.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
NM_001123369.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
NM_001123369.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
NM_001123369.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
NM_002721.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
NM_002721.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
NM_002721.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
NM_002721.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
NM_002721.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
NM_002721.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
NM_002721.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
NM_002721.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
NM_002721.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
NM_002721.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
NM_002721.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
NM_002721.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011519.hg.1 |
NM_002721.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09019948.hg.1 |
TC09001588.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09019948.hg.1 |
ENST00000373546.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09019948.hg.1 |
ENST00000373547.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09019948.hg.1 |
ENST00000415905.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09019948.hg.1 |
ENST00000451402.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09019948.hg.1 |
NM_001123355.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09019948.hg.1 |
NM_001123369.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09019948.hg.1 |
NM_002721.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
ENST00000373547.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
ENST00000373547.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
ENST00000373547.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
ENST00000373547.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
ENST00000373547.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
ENST00000373547.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
ENST00000373547.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
ENST00000373547.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
ENST00000373547.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
ENST00000373547.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
ENST00000373547.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
ENST00000373547.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
ENST00000415905.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
ENST00000415905.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
ENST00000415905.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
ENST00000415905.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
ENST00000415905.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
ENST00000415905.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
ENST00000415905.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
ENST00000415905.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
ENST00000415905.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
ENST00000415905.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
ENST00000415905.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
ENST00000415905.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
ENST00000451402.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
ENST00000451402.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
ENST00000451402.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
ENST00000451402.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
ENST00000451402.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
ENST00000451402.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
ENST00000451402.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
ENST00000451402.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
ENST00000451402.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
ENST00000451402.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
ENST00000451402.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
ENST00000451402.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
ENST00000451402.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
ENST00000451402.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
ENST00000451402.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
ENST00000456642.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
ENST00000456642.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
ENST00000456642.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
ENST00000456642.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
ENST00000456642.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
ENST00000456642.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
ENST00000456642.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
NM_001123355.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
NM_001123355.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
NM_001123355.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
NM_001123355.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
NM_001123355.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
NM_001123355.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
NM_001123355.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
NM_001123355.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
NM_001123355.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
NM_001123355.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
NM_001123355.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
NM_001123355.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
NM_001123355.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
NM_001123355.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
NM_001123355.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
NM_001123369.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
NM_001123369.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
NM_001123369.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
NM_001123369.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
NM_001123369.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
NM_001123369.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
NM_001123369.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
NM_001123369.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
NM_001123369.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
NM_001123369.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
NM_001123369.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
NM_001123369.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
NM_002721.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
NM_002721.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
NM_002721.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
NM_002721.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
NM_002721.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
NM_002721.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
NM_002721.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
NM_002721.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
NM_002721.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
NM_002721.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
NM_002721.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
NM_002721.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011519.hg.1 |
NM_002721.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900178583.hg.1 |
ENST00000373547.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900178583.hg.1 |
ENST00000415905.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900178583.hg.1 |
ENST00000451402.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900178583.hg.1 |
NM_001123355.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900178583.hg.1 |
NM_001123369.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900178583.hg.1 |
NM_002721.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900178583.hg.1 |
ENST00000373547.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900178583.hg.1 |
ENST00000415905.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900178583.hg.1 |
ENST00000451402.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900178583.hg.1 |
NM_001123355.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900178583.hg.1 |
NM_001123369.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900178583.hg.1 |
NM_002721.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900178583.hg.1 |
TC0900011519.hg.1 |
Clariom D TC |