| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000372770.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000372770.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000372770.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000372770.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000372770.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000372770.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000372770.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000372770.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000372770.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000372770.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000372770.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000372770.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000372770.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000372770.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000372770.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000372770.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000372770.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000459581.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000494417.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000494417.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000494417.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000494417.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000494417.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000494417.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001499.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001499.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001499.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001499.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001499.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001499.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001499.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001499.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001499.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001499.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001499.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001499.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001499.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001499.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001499.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001499.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001499.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001499.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09008458.hg.1 |
TC09000693.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09008458.hg.1 |
ENST00000309971.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09008458.hg.1 |
ENST00000372770.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09008458.hg.1 |
ENST00000539582.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09008458.hg.1 |
NM_001003722.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09008458.hg.1 |
NM_001499.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900184768.hg.1 |
ENST00000309971.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900184768.hg.1 |
ENST00000372770.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900184768.hg.1 |
NM_001003722.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900184768.hg.1 |
NM_001499.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900184768.hg.1 |
ENST00000309971.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900184768.hg.1 |
ENST00000372770.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900184768.hg.1 |
NM_001003722.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900184768.hg.1 |
NM_001499.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900184768.hg.1 |
TC0900012178.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000372770.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000372770.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000372770.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000372770.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000372770.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000372770.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000372770.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000372770.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000372770.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000372770.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000372770.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000372770.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000372770.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000372770.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000372770.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000372770.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000372770.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000459581.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000494417.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000494417.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000494417.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000494417.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000494417.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000494417.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001499.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001499.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001499.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001499.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001499.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001499.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001499.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001499.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001499.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001499.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001499.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001499.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001499.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001499.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001499.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001499.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001499.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001499.2 |
Refseq |