Assay Details
Gene Symbol: KCNJ3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_002239.3 | NP_002230.1 | 2 - 3 | 1111 | 106 | |
GenBank mRNA | AK293924.1 | - | 2 - 3 | 259 | 106 | |
BC022495.1 | - | 2 - 3 | 946 | 4817322 | 106 | |
U50964.1 | - | 2 - 3 | 1112 | 106 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC02000947.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR02013866.hg.1 | TC02000947.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02013866.hg.1 | ENST00000295101.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02013866.hg.1 | NM_002239.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02013864.hg.1 | TC02000947.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02013864.hg.1 | ENST00000295101.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02013864.hg.1 | ENST00000493505.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02013864.hg.1 | ENST00000544049.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02013864.hg.1 | NM_002239.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009707.hg.1 | ENST00000295101.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009707.hg.1 | ENST00000295101.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009707.hg.1 | ENST00000295101.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009707.hg.1 | ENST00000295101.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009707.hg.1 | ENST00000295101.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009707.hg.1 | ENST00000295101.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009707.hg.1 | ENST00000295101.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009707.hg.1 | ENST00000295101.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009707.hg.1 | ENST00000295101.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009707.hg.1 | ENST00000493505.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009707.hg.1 | ENST00000493505.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009707.hg.1 | ENST00000493505.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009707.hg.1 | ENST00000544049.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009707.hg.1 | ENST00000544049.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009707.hg.1 | ENST00000544049.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009707.hg.1 | ENST00000544049.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009707.hg.1 | ENST00000544049.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009707.hg.1 | ENST00000544049.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009707.hg.1 | ENST00000544049.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009707.hg.1 | NM_001260508.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009707.hg.1 | NM_001260508.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009707.hg.1 | NM_001260508.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009707.hg.1 | NM_001260508.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009707.hg.1 | NM_001260508.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009707.hg.1 | NM_001260508.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009707.hg.1 | NM_001260508.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009707.hg.1 | NM_001260509.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009707.hg.1 | NM_001260509.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009707.hg.1 | NM_001260509.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009707.hg.1 | NM_001260509.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009707.hg.1 | NM_001260509.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009707.hg.1 | NM_001260510.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009707.hg.1 | NM_001260510.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009707.hg.1 | NM_001260510.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009707.hg.1 | NM_001260510.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009707.hg.1 | NM_002239.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009707.hg.1 | NM_002239.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009707.hg.1 | NM_002239.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009707.hg.1 | NM_002239.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009707.hg.1 | NM_002239.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009707.hg.1 | NM_002239.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009707.hg.1 | NM_002239.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009707.hg.1 | NM_002239.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02013868.hg.1 | TC02000947.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02013868.hg.1 | ENST00000295101.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02013868.hg.1 | NM_002239.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200167795.hg.1 | ENST00000295101.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200167795.hg.1 | ENST00000544049.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200167795.hg.1 | NM_001260508.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200167795.hg.1 | NM_002239.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200167795.hg.1 | ENST00000295101.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200167795.hg.1 | ENST00000544049.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200167795.hg.1 | NM_001260508.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200167795.hg.1 | NM_002239.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200167795.hg.1 | TC0200009707.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0200009707.hg.1 | ENST00000295101.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009707.hg.1 | ENST00000295101.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009707.hg.1 | ENST00000295101.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009707.hg.1 | ENST00000295101.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009707.hg.1 | ENST00000295101.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009707.hg.1 | ENST00000295101.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009707.hg.1 | ENST00000295101.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009707.hg.1 | ENST00000295101.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009707.hg.1 | ENST00000295101.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009707.hg.1 | ENST00000493505.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009707.hg.1 | ENST00000493505.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009707.hg.1 | ENST00000493505.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009707.hg.1 | ENST00000544049.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009707.hg.1 | ENST00000544049.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009707.hg.1 | ENST00000544049.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009707.hg.1 | ENST00000544049.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009707.hg.1 | ENST00000544049.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009707.hg.1 | ENST00000544049.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009707.hg.1 | ENST00000544049.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009707.hg.1 | NM_001260508.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009707.hg.1 | NM_001260508.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009707.hg.1 | NM_001260508.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009707.hg.1 | NM_001260508.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009707.hg.1 | NM_001260508.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009707.hg.1 | NM_001260508.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009707.hg.1 | NM_001260508.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009707.hg.1 | NM_001260509.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009707.hg.1 | NM_001260509.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009707.hg.1 | NM_001260509.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009707.hg.1 | NM_001260509.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009707.hg.1 | NM_001260509.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009707.hg.1 | NM_001260510.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009707.hg.1 | NM_001260510.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009707.hg.1 | NM_001260510.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009707.hg.1 | NM_001260510.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009707.hg.1 | NM_002239.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009707.hg.1 | NM_002239.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009707.hg.1 | NM_002239.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009707.hg.1 | NM_002239.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009707.hg.1 | NM_002239.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009707.hg.1 | NM_002239.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009707.hg.1 | NM_002239.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009707.hg.1 | NM_002239.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200167793.hg.1 | ENST00000295101.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200167793.hg.1 | ENST00000493505.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200167793.hg.1 | ENST00000544049.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200167793.hg.1 | NM_001260508.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200167793.hg.1 | NM_002239.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200167793.hg.1 | ENST00000295101.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200167793.hg.1 | ENST00000493505.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200167793.hg.1 | ENST00000544049.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200167793.hg.1 | NM_001260508.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200167793.hg.1 | NM_002239.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200167793.hg.1 | TC0200009707.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0200167794.hg.1 | ENST00000295101.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200167794.hg.1 | ENST00000544049.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200167794.hg.1 | NM_001260508.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200167794.hg.1 | NM_002239.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200167794.hg.1 | ENST00000295101.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200167794.hg.1 | ENST00000544049.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200167794.hg.1 | NM_001260508.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200167794.hg.1 | NM_002239.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200167794.hg.1 | TC0200009707.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 120159 | - | - |
ABI Human Genome Survey Microarray Version 2.0 | 300784 | - | - |
