Assay Details
| Gene Symbol: PKLR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181871.3 | NP_870986.1 | 1 - 2 | 121 | 66 | |
| GenBank mRNA | AK225947.1 | - | 1 - 2 | 121 | 66 | |
| M15465.1 | - | 1 - 2 | 75 | 66 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01003312.hg
Additional Information:
This assay is in the 5'UTR region of the M15465.1,NM_181871.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
response to hypoxia
ATP biosynthetic process
response to nutrient
response to heat
response to glucose
response to lithium ion
cellular response to insulin stimulus
response to ATP
pyruvate biosynthetic process
response to cAMP
canonical glycolysis
magnesium ion binding
pyruvate kinase activity
ATP binding
kinase activity
potassium ion binding
response to hypoxia
ATP biosynthetic process
response to nutrient
response to heat
response to glucose
response to lithium ion
cellular response to insulin stimulus
response to ATP
pyruvate biosynthetic process
response to cAMP
canonical glycolysis
magnesium ion binding
pyruvate kinase activity
ATP binding
kinase activity
potassium ion binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01051422.hg.1 | TC01003312.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01051422.hg.1 | ENST00000392414.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01051422.hg.1 | NM_181871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015953.hg.1 | ENST00000342741.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015953.hg.1 | ENST00000342741.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015953.hg.1 | ENST00000342741.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015953.hg.1 | ENST00000342741.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015953.hg.1 | ENST00000342741.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015953.hg.1 | ENST00000342741.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015953.hg.1 | ENST00000342741.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015953.hg.1 | ENST00000342741.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015953.hg.1 | ENST00000342741.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015953.hg.1 | ENST00000342741.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015953.hg.1 | ENST00000342741.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015953.hg.1 | ENST00000342741.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015953.hg.1 | ENST00000342741.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015953.hg.1 | ENST00000342741.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015953.hg.1 | ENST00000392414.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015953.hg.1 | ENST00000392414.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015953.hg.1 | ENST00000392414.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015953.hg.1 | ENST00000392414.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015953.hg.1 | ENST00000392414.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015953.hg.1 | ENST00000392414.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015953.hg.1 | ENST00000392414.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015953.hg.1 | ENST00000392414.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015953.hg.1 | ENST00000392414.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015953.hg.1 | ENST00000392414.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015953.hg.1 | ENST00000392414.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015953.hg.1 | ENST00000392414.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015953.hg.1 | ENST00000392414.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015953.hg.1 | ENST00000392414.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015953.hg.1 | ENST00000392414.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015953.hg.1 | ENST00000434082.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015953.hg.1 | ENST00000434082.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015953.hg.1 | ENST00000434082.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015953.hg.1 | ENST00000434082.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015953.hg.1 | ENST00000434082.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015953.hg.1 | NM_000298.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015953.hg.1 | NM_000298.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015953.hg.1 | NM_000298.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015953.hg.1 | NM_000298.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015953.hg.1 | NM_000298.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015953.hg.1 | NM_000298.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015953.hg.1 | NM_000298.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015953.hg.1 | NM_000298.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015953.hg.1 | NM_000298.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015953.hg.1 | NM_000298.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015953.hg.1 | NM_000298.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015953.hg.1 | NM_000298.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015953.hg.1 | NM_000298.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015953.hg.1 | NM_000298.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015953.hg.1 | NM_000298.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015953.hg.1 | NM_000298.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015953.hg.1 | NM_181871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015953.hg.1 | NM_181871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015953.hg.1 | NM_181871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015953.hg.1 | NM_181871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015953.hg.1 | NM_181871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015953.hg.1 | NM_181871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015953.hg.1 | NM_181871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015953.hg.1 | NM_181871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015953.hg.1 | NM_181871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015953.hg.1 | NM_181871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015953.hg.1 | NM_181871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015953.hg.1 | NM_181871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015953.hg.1 | NM_181871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015953.hg.1 | NM_181871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015953.hg.1 | NM_181871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015953.hg.1 | NM_181871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01051420.hg.1 | TC01003312.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01051420.hg.1 | ENST00000271946.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01051420.hg.1 | ENST00000392414.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01051420.hg.1 | ENST00000423816.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01051420.hg.1 | NM_181871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015953.hg.1 | ENST00000342741.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015953.hg.1 | ENST00000342741.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015953.hg.1 | ENST00000342741.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015953.hg.1 | ENST00000342741.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015953.hg.1 | ENST00000342741.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015953.hg.1 | ENST00000342741.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015953.hg.1 | ENST00000342741.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015953.hg.1 | ENST00000342741.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015953.hg.1 | ENST00000342741.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015953.hg.1 | ENST00000342741.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015953.hg.1 | ENST00000342741.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015953.hg.1 | ENST00000342741.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015953.hg.1 | ENST00000342741.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015953.hg.1 | ENST00000342741.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015953.hg.1 | ENST00000392414.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015953.hg.1 | ENST00000392414.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015953.hg.1 | ENST00000392414.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015953.hg.1 | ENST00000392414.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015953.hg.1 | ENST00000392414.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015953.hg.1 | ENST00000392414.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015953.hg.1 | ENST00000392414.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015953.hg.1 | ENST00000392414.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015953.hg.1 | ENST00000392414.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015953.hg.1 | ENST00000392414.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015953.hg.1 | ENST00000392414.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015953.hg.1 | ENST00000392414.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015953.hg.1 | ENST00000392414.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015953.hg.1 | ENST00000392414.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015953.hg.1 | ENST00000392414.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015953.hg.1 | ENST00000434082.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015953.hg.1 | ENST00000434082.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015953.hg.1 | ENST00000434082.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015953.hg.1 | ENST00000434082.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015953.hg.1 | ENST00000434082.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015953.hg.1 | NM_000298.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015953.hg.1 | NM_000298.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015953.hg.1 | NM_000298.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015953.hg.1 | NM_000298.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015953.hg.1 | NM_000298.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015953.hg.1 | NM_000298.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015953.hg.1 | NM_000298.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015953.hg.1 | NM_000298.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015953.hg.1 | NM_000298.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015953.hg.1 | NM_000298.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015953.hg.1 | NM_000298.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015953.hg.1 | NM_000298.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015953.hg.1 | NM_000298.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015953.hg.1 | NM_000298.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015953.hg.1 | NM_000298.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015953.hg.1 | NM_000298.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015953.hg.1 | NM_181871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015953.hg.1 | NM_181871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015953.hg.1 | NM_181871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015953.hg.1 | NM_181871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015953.hg.1 | NM_181871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015953.hg.1 | NM_181871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015953.hg.1 | NM_181871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015953.hg.1 | NM_181871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015953.hg.1 | NM_181871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015953.hg.1 | NM_181871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015953.hg.1 | NM_181871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015953.hg.1 | NM_181871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015953.hg.1 | NM_181871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015953.hg.1 | NM_181871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015953.hg.1 | NM_181871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015953.hg.1 | NM_181871.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100220483.hg.1 | ENST00000392414.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100220483.hg.1 | NM_181871.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100220483.hg.1 | ENST00000392414.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100220483.hg.1 | NM_181871.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100220483.hg.1 | TC0100015953.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100220482.hg.1 | ENST00000392414.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100220482.hg.1 | ENST00000434082.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100220482.hg.1 | NM_181871.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100220482.hg.1 | ENST00000392414.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100220482.hg.1 | ENST00000434082.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100220482.hg.1 | NM_181871.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100220482.hg.1 | TC0100015953.hg.1 | Clariom D TC |

PKLR
Genomic Map