Assay Details
| Gene Symbol: ENOPH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001292017.1 | NP_001278946.1 | 3 - 4 | 712 | 68 | |
| NM_021204.4 | NP_067027.1 | 3 - 4 | 741 | 68 | ||
| GenBank mRNA | AF087881.1 | - | 3 - 4 | 611 | 68 | |
| AF113125.1 | - | 3 - 4 | 612 | 68 | ||
| AF177286.1 | - | 1 - 2 | 178 | 68 | ||
| AK022656.1 | - | 3 - 4 | 607 | 68 | ||
| BC065815.1 | - | 3 - 4 | 579 | 6526329 | 68 | |
| CN335976.1 | - | 3 - 4 | 553 | 68 | ||
| CR457141.1 | - | 3 - 4 | 391 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

ENOPH1
Genomic Map