Assay Details
Gene Symbol: PNP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000270.3 | NP_000261.2 | 3 - 4 | 435 | 75 | |
GenBank mRNA | AK313490.1 | - | 3 - 4 | 408 | 75 | |
BC104206.1 | - | 3 - 4 | 376 | 40031416 | 75 | |
BC104207.1 | - | 3 - 4 | 376 | 40031417 | 75 | |
BC106074.1 | - | 3 - 4 | 431 | 3859664 | 75 | |
CR407607.1 | - | 2 - 3 | 289 | 75 | ||
EU794649.1 | - | 3 - 4 | 431 | 75 | ||
X00737.1 | - | 3 - 4 | 398 | 75 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC14000056.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
nucleobase-containing compound metabolic process
inosine catabolic process
purine nucleotide catabolic process
nicotinamide riboside catabolic process
immune response
NAD biosynthesis via nicotinamide riboside salvage pathway
urate biosynthetic process
positive regulation of T cell proliferation
response to drug
purine-containing compound salvage
positive regulation of alpha-beta T cell differentiation
interleukin-2 secretion
nucleoside binding
purine nucleobase binding
purine-nucleoside phosphorylase activity
drug binding
phosphate ion binding
nucleobase-containing compound metabolic process
inosine catabolic process
purine nucleotide catabolic process
nicotinamide riboside catabolic process
immune response
NAD biosynthesis via nicotinamide riboside salvage pathway
urate biosynthetic process
positive regulation of T cell proliferation
response to drug
purine-containing compound salvage
positive regulation of alpha-beta T cell differentiation
interleukin-2 secretion
nucleoside binding
purine nucleobase binding
purine-nucleoside phosphorylase activity
drug binding
phosphate ion binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR14000251.hg.1 | ENST00000361505.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR14000251.hg.1 | NM_000270.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR14000251.hg.1 | TC14000056.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000361505.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000361505.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000361505.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000361505.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000361505.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000361505.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000361505.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000361505.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000361505.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000361505.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000361505.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000361505.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000553418.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000553418.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000553418.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000553418.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000553418.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000553591.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000553591.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000553591.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000553591.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000553591.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000553591.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000554056.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000554056.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000554056.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000554056.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000554056.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000554056.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000554056.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000554056.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000554056.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000554056.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000554056.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000554056.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000554065.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000554065.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000554065.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000554065.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000556293.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000556293.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000556293.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000556754.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000556754.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000556754.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000556754.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000556754.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000556754.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000556754.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000556754.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000556754.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000556754.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000556754.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000556754.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000556754.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000557229.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000557229.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000557229.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000557229.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000557229.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000557229.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000557229.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000557229.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | NM_000270.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | NM_000270.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | NM_000270.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | NM_000270.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | NM_000270.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | NM_000270.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | NM_000270.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | NM_000270.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | NM_000270.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | NM_000270.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | NM_000270.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | NM_000270.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | NM_000270.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000361505.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000361505.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000361505.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000361505.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000361505.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000361505.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000361505.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000361505.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000361505.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000361505.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000361505.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000361505.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000553418.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000553418.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000553418.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000553418.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000553418.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000553591.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000553591.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000553591.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000553591.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000553591.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000553591.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000554056.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000554056.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000554056.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000554056.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000554056.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000554056.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000554056.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000554056.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000554056.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000554056.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000554056.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000554056.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000554065.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000554065.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000554065.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000554065.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000556293.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000556293.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000556293.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000556754.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000556754.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000556754.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000556754.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000556754.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000556754.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000556754.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000556754.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000556754.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000556754.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000556754.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000556754.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000556754.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000557229.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000557229.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000557229.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000557229.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000557229.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000557229.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000557229.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000557229.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | NM_000270.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | NM_000270.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | NM_000270.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | NM_000270.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | NM_000270.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | NM_000270.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | NM_000270.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | NM_000270.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | NM_000270.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | NM_000270.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | NM_000270.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | NM_000270.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | NM_000270.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400144495.hg.1 | ENST00000361505.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400144495.hg.1 | ENST00000553591.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400144495.hg.1 | ENST00000554056.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400144495.hg.1 | ENST00000554065.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400144495.hg.1 | ENST00000556754.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400144495.hg.1 | ENST00000557229.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400144495.hg.1 | NM_000270.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400144495.hg.1 | ENST00000361505.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400144495.hg.1 | ENST00000553591.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400144495.hg.1 | ENST00000554056.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400144495.hg.1 | ENST00000554065.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400144495.hg.1 | ENST00000556754.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400144495.hg.1 | ENST00000557229.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400144495.hg.1 | NM_000270.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400144495.hg.1 | TC1400006521.hg.1 | Clariom D TC |
