Assay Details
Gene Symbol: HLCS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000411.6 | NP_000402.3 | 3 - 4 | 361 | 76 | |
NM_001242785.1 | NP_001229714.1 | 3 - 4 | 454 | 76 | ||
XM_006723995.1 | XP_006724058.1 | 3 - 4 | 486 | 76 | ||
XM_011529538.1 | XP_011527840.1 | 3 - 4 | 341 | 76 | ||
GenBank mRNA | AK314189.1 | - | 2 - 3 | 228 | 76 | |
BC060787.1 | - | 3 - 4 | 319 | 30343493 | 76 | |
D23672.2 | - | 3 - 4 | 352 | 76 | ||
DA334377.1 | - | 3 - 4 | 361 | 76 | ||
DB290499.1 | - | 3 - 4 | 486 | 76 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK314189.1, BC060787.1, D23672.2, NM_000411.6, NM_001242785.1, XM_006723995.1, XM_011529538.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
