Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004376.hg.1 |
ENST00000298249.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004376.hg.1 |
ENST00000373758.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004376.hg.1 |
ENST00000438249.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004376.hg.1 |
NM_001001330.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004376.hg.1 |
TC10000389.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007797.hg.1 |
ENST00000298249.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007797.hg.1 |
ENST00000298249.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007797.hg.1 |
ENST00000298249.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007797.hg.1 |
ENST00000298249.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007797.hg.1 |
ENST00000298249.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007797.hg.1 |
ENST00000298249.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007797.hg.1 |
ENST00000298249.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007797.hg.1 |
ENST00000298249.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007797.hg.1 |
ENST00000298249.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007797.hg.1 |
ENST00000298249.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007797.hg.1 |
ENST00000298249.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007797.hg.1 |
ENST00000298249.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007797.hg.1 |
ENST00000373758.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007797.hg.1 |
ENST00000373758.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007797.hg.1 |
ENST00000373758.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007797.hg.1 |
ENST00000373758.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007797.hg.1 |
ENST00000373758.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007797.hg.1 |
ENST00000373758.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007797.hg.1 |
ENST00000373758.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007797.hg.1 |
ENST00000373758.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007797.hg.1 |
ENST00000373758.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007797.hg.1 |
ENST00000373758.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007797.hg.1 |
ENST00000373758.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007797.hg.1 |
ENST00000373758.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007797.hg.1 |
NM_001001330.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007797.hg.1 |
NM_001001330.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007797.hg.1 |
NM_001001330.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007797.hg.1 |
NM_001001330.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007797.hg.1 |
NM_001001330.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007797.hg.1 |
NM_001001330.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007797.hg.1 |
NM_001001330.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007797.hg.1 |
NM_001001330.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007797.hg.1 |
NM_001001330.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007797.hg.1 |
NM_001001330.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007797.hg.1 |
NM_001001330.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007797.hg.1 |
NM_001001330.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007797.hg.1 |
ENST00000298249.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007797.hg.1 |
ENST00000298249.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007797.hg.1 |
ENST00000298249.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007797.hg.1 |
ENST00000298249.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007797.hg.1 |
ENST00000298249.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007797.hg.1 |
ENST00000298249.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007797.hg.1 |
ENST00000298249.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007797.hg.1 |
ENST00000298249.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007797.hg.1 |
ENST00000298249.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007797.hg.1 |
ENST00000298249.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007797.hg.1 |
ENST00000298249.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007797.hg.1 |
ENST00000298249.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007797.hg.1 |
ENST00000373758.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007797.hg.1 |
ENST00000373758.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007797.hg.1 |
ENST00000373758.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007797.hg.1 |
ENST00000373758.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007797.hg.1 |
ENST00000373758.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007797.hg.1 |
ENST00000373758.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007797.hg.1 |
ENST00000373758.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007797.hg.1 |
ENST00000373758.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007797.hg.1 |
ENST00000373758.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007797.hg.1 |
ENST00000373758.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007797.hg.1 |
ENST00000373758.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007797.hg.1 |
ENST00000373758.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007797.hg.1 |
NM_001001330.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007797.hg.1 |
NM_001001330.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007797.hg.1 |
NM_001001330.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007797.hg.1 |
NM_001001330.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007797.hg.1 |
NM_001001330.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007797.hg.1 |
NM_001001330.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007797.hg.1 |
NM_001001330.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007797.hg.1 |
NM_001001330.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007797.hg.1 |
NM_001001330.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007797.hg.1 |
NM_001001330.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007797.hg.1 |
NM_001001330.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007797.hg.1 |
NM_001001330.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000152784.hg.1 |
ENST00000298249.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000152784.hg.1 |
ENST00000373758.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000152784.hg.1 |
NM_001001330.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000152784.hg.1 |
ENST00000298249.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000152784.hg.1 |
ENST00000373758.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000152784.hg.1 |
NM_001001330.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000152784.hg.1 |
TC1000007797.hg.1 |
Clariom D TC |