Assay Details
| Gene Symbol: SYNGR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320523.1 | NP_001307452.1 | 2 - 3 | 407 | 84 | |
| GenBank mRNA | CF122406.1 | - | 2 - 3 | 338 | 84 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | ENST00000225777.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | ENST00000225777.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | ENST00000225777.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | ENST00000225777.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | ENST00000225777.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | ENST00000225777.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | ENST00000225777.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | ENST00000225777.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | ENST00000225777.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | ENST00000225777.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | ENST00000225777.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | ENST00000225777.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | ENST00000585591.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | ENST00000585591.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | ENST00000585591.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | ENST00000585591.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | ENST00000585591.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | ENST00000585591.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | ENST00000588282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | ENST00000588282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | ENST00000588282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | ENST00000588282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | ENST00000588282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | ENST00000588282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | ENST00000588282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | ENST00000588282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | ENST00000589168.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | ENST00000589168.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | ENST00000589168.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | ENST00000589168.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | ENST00000589168.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | ENST00000589168.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | ENST00000589168.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | ENST00000589168.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | ENST00000589168.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | ENST00000589183.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | ENST00000589183.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | ENST00000589183.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | ENST00000589183.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | ENST00000589183.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | ENST00000589711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | ENST00000589711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | ENST00000589711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | ENST00000589711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | ENST00000589711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | ENST00000589711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | ENST00000589711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | ENST00000589711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | ENST00000589711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | ENST00000589711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | ENST00000590201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | ENST00000590201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | ENST00000590201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | ENST00000590201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | ENST00000590201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | ENST00000590201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | ENST00000590201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | ENST00000590201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | ENST00000590201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | ENST00000590201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | ENST00000590201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | ENST00000591770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | ENST00000591770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | ENST00000591770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | ENST00000591770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | ENST00000592456.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | ENST00000592456.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | ENST00000592456.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | ENST00000592456.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | ENST00000592456.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | ENST00000592456.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | ENST00000592456.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | ENST00000592456.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | ENST00000592456.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | ENST00000592456.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | NM_004710.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | NM_004710.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | NM_004710.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | NM_004710.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | NM_004710.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | NM_004710.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | NM_004710.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009040.hg.1 | NM_004710.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | ENST00000225777.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | ENST00000225777.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | ENST00000225777.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | ENST00000225777.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | ENST00000225777.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | ENST00000225777.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | ENST00000225777.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | ENST00000225777.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | ENST00000225777.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | ENST00000225777.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | ENST00000225777.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | ENST00000225777.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | ENST00000585591.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | ENST00000585591.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | ENST00000585591.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | ENST00000585591.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | ENST00000585591.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | ENST00000585591.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | ENST00000588282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | ENST00000588282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | ENST00000588282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | ENST00000588282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | ENST00000588282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | ENST00000588282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | ENST00000588282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | ENST00000588282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | ENST00000589168.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | ENST00000589168.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | ENST00000589168.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | ENST00000589168.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | ENST00000589168.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | ENST00000589168.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | ENST00000589168.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | ENST00000589168.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | ENST00000589168.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | ENST00000589183.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | ENST00000589183.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | ENST00000589183.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | ENST00000589183.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | ENST00000589183.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | ENST00000589711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | ENST00000589711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | ENST00000589711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | ENST00000589711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | ENST00000589711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | ENST00000589711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | ENST00000589711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | ENST00000589711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | ENST00000589711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | ENST00000589711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | ENST00000590201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | ENST00000590201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | ENST00000590201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | ENST00000590201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | ENST00000590201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | ENST00000590201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | ENST00000590201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | ENST00000590201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | ENST00000590201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | ENST00000590201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | ENST00000590201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | ENST00000591770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | ENST00000591770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | ENST00000591770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | ENST00000591770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | ENST00000592456.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | ENST00000592456.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | ENST00000592456.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | ENST00000592456.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | ENST00000592456.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | ENST00000592456.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | ENST00000592456.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | ENST00000592456.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | ENST00000592456.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | ENST00000592456.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | NM_004710.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | NM_004710.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | NM_004710.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | NM_004710.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | NM_004710.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | NM_004710.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | NM_004710.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009040.hg.1 | NM_004710.4 | Refseq |

SYNGR2
Genomic Map