Assay Details
| Gene Symbol: TRIM41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033549.4 | NP_291027.3 | 3 - 4 | 1878 | 134 | |
| NM_201627.2 | NP_963921.1 | 3 - 4 | 1878 | 134 | ||
| XM_006714929.2 | XP_006714992.1 | 3 - 4 | 1876 | 134 | ||
| XM_006714930.2 | XP_006714993.1 | 3 - 4 | 1876 | 134 | ||
| XM_006714931.3 | XP_006714994.1 | 3 - 4 | 1876 | 134 | ||
| XM_006714932.3 | XP_006714995.1 | 3 - 4 | 1876 | 134 | ||
| GenBank mRNA | AB063180.1 | - | 3 - 4 | 904 | 134 | |
| AB100366.1 | - | 3 - 4 | 1141 | 134 | ||
| AB100367.1 | - | 3 - 4 | 1141 | 134 | ||
| AK027601.1 | - | 3 - 4 | 1851 | 134 | ||
| AX775789.1 | - | 3 - 4 | 617 | 134 | ||
| AX775791.1 | - | 3 - 4 | 617 | 134 | ||
| AX775793.1 | - | 3 - 4 | 1851 | 134 | ||
| AX775797.1 | - | 3 - 4 | 909 | 134 | ||
| AX775799.1 | - | 3 - 4 | 1851 | 134 | ||
| BC018765.1 | - | 3 - 4 | 909 | 134 | ||
| BC090953.1 | - | 3 - 4 | 1421 | 6017628 | 134 | |
More Information
Set Membership: |
|

TRIM41
Genomic Map