Assay Details
| Gene Symbol: CYP2S1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK296087.1 | - | 2 - 3 | 366 | 73 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK296087.1 transcripts.
Set Membership: |
|

CYP2S1
Genomic Map