Assay Details
| Gene Symbol: HAO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005783.2 | NP_001005783.2 | 7 - 8 | 1289 | 120 | |
| NM_016527.3 | NP_057611.1 | 6 - 7 | 1069 | 120 | ||
| XM_011541560.1 | XP_011539862.1 | 4 - 5 | 732 | 120 | ||
| GenBank mRNA | AF231917.1 | - | 6 - 7 | 1007 | 120 | |
| AK298289.1 | - | 7 - 8 | 1289 | 120 | ||
| AY513277.1 | - | 6 - 7 | 974 | 120 | ||
| BC020863.1 | - | 7 - 8 | 1256 | 4557798 | 120 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01001037.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
fatty acid oxidation
cellular lipid metabolic process
(S)-2-hydroxy-acid oxidase activity
receptor binding
FMN binding
very-long-chain-(S)-2-hydroxy-acid oxidase activity
long-chain-(S)-2-hydroxy-long-chain-acid oxidase activity
medium-chain-(S)-2-hydroxy-acid oxidase activity
fatty acid oxidation
cellular lipid metabolic process
(S)-2-hydroxy-acid oxidase activity
receptor binding
FMN binding
very-long-chain-(S)-2-hydroxy-acid oxidase activity
long-chain-(S)-2-hydroxy-long-chain-acid oxidase activity
medium-chain-(S)-2-hydroxy-acid oxidase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01016452.hg.1 | TC01001037.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01016452.hg.1 | ENST00000325945.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01016452.hg.1 | ENST00000361035.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01016452.hg.1 | ENST00000482991.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01016452.hg.1 | NM_001005783.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01016452.hg.1 | NM_016527.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000325945.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000325945.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000325945.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000325945.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000325945.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000325945.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000325945.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000325945.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000325945.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000325945.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000325945.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000361035.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000361035.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000361035.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000361035.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000361035.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000361035.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000361035.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000361035.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000361035.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000361035.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000361035.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000361035.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000361035.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000457318.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000457318.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000457318.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000457318.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000457318.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000457318.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000457318.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000482991.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000482991.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000482991.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000482991.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000622548.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000622548.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000622548.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000622548.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000622548.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000622548.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000622548.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000622548.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000622548.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000622548.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000622548.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000622548.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000622548.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000622548.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | NM_001005783.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | NM_001005783.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | NM_001005783.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | NM_001005783.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | NM_001005783.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | NM_001005783.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | NM_001005783.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | NM_001005783.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | NM_001005783.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | NM_001005783.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | NM_001005783.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | NM_001005783.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | NM_001005783.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | NM_001005783.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | NM_016527.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | NM_016527.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | NM_016527.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | NM_016527.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | NM_016527.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | NM_016527.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | NM_016527.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | NM_016527.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | NM_016527.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | NM_016527.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | NM_016527.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | NM_016527.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | NM_016527.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000325945.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000325945.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000325945.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000325945.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000325945.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000325945.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000325945.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000325945.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000325945.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000325945.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000325945.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000361035.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000361035.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000361035.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000361035.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000361035.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000361035.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000361035.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000361035.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000361035.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000361035.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000361035.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000361035.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000361035.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000457318.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000457318.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000457318.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000457318.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000457318.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000457318.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000457318.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000482991.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000482991.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000482991.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000482991.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000622548.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000622548.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000622548.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000622548.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000622548.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000622548.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000622548.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000622548.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000622548.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000622548.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000622548.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000622548.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000622548.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000622548.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | NM_001005783.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | NM_001005783.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | NM_001005783.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | NM_001005783.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | NM_001005783.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | NM_001005783.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | NM_001005783.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | NM_001005783.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | NM_001005783.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | NM_001005783.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | NM_001005783.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | NM_001005783.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | NM_001005783.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | NM_001005783.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | NM_016527.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | NM_016527.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | NM_016527.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | NM_016527.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | NM_016527.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | NM_016527.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | NM_016527.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | NM_016527.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | NM_016527.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | NM_016527.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | NM_016527.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | NM_016527.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | NM_016527.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100169148.hg.1 | ENST00000325945.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100169148.hg.1 | ENST00000361035.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100169148.hg.1 | ENST00000482991.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100169148.hg.1 | ENST00000622548.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100169148.hg.1 | NM_001005783.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100169148.hg.1 | NM_016527.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100169148.hg.1 | ENST00000325945.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100169148.hg.1 | ENST00000361035.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100169148.hg.1 | ENST00000482991.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100169148.hg.1 | ENST00000622548.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100169148.hg.1 | NM_001005783.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100169148.hg.1 | NM_016527.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100169148.hg.1 | TC0100009608.hg.1 | Clariom D TC |

HAO2
Genomic Map