Assay Details
Gene Symbol: PPP2CA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_002715.2 | NP_002706.1 | 2 - 3 | 713 | 146 | |
GenBank mRNA | AB451240.1 | - | 2 - 3 | 315 | 146 | |
AB451365.1 | - | 2 - 3 | 315 | 146 | ||
AK057424.1 | - | 2 - 3 | 445 | 146 | ||
AK097599.1 | - | 2 - 3 | 525 | 146 | ||
BC000400.2 | - | 2 - 3 | 514 | 2822635 | 146 | |
BC002657.2 | - | 2 - 3 | 514 | 3607862 | 146 | |
BC019275.1 | - | 2 - 3 | 514 | 2822635 | 146 | |
BC031696.1 | - | 2 - 3 | 514 | 5001649 | 146 | |
CR457417.1 | - | 2 - 3 | 315 | 146 | ||
J03804.1 | - | 2 - 3 | 497 | 146 | ||
X12646.1 | - | 2 - 3 | 398 | 146 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC05003406.hg.1, TC05003407.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
inactivation of MAPK activity
regulation of DNA replication
regulation of transcription, DNA-templated
protein dephosphorylation
ceramide metabolic process
apoptotic process
mitotic nuclear envelope reassembly
mesoderm development
RNA splicing
response to organic substance
negative regulation of epithelial to mesenchymal transition
second-messenger-mediated signaling
regulation of Wnt signaling pathway
regulation of cell adhesion
negative regulation of cell growth
regulation of growth
negative regulation of tyrosine phosphorylation of Stat3 protein
regulation of cell differentiation
meiotic cell cycle
positive regulation of protein serine/threonine kinase activity
phosphoprotein phosphatase activity
protein binding
protein C-terminus binding
metal ion binding
GABA receptor binding
nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
inactivation of MAPK activity
regulation of DNA replication
regulation of transcription, DNA-templated
protein dephosphorylation
ceramide metabolic process
apoptotic process
mitotic nuclear envelope reassembly
mesoderm development
RNA splicing
response to organic substance
negative regulation of epithelial to mesenchymal transition
second-messenger-mediated signaling
regulation of Wnt signaling pathway
regulation of cell adhesion
negative regulation of cell growth
regulation of growth
negative regulation of tyrosine phosphorylation of Stat3 protein
regulation of cell differentiation
meiotic cell cycle
positive regulation of protein serine/threonine kinase activity
phosphoprotein phosphatase activity
protein binding
protein C-terminus binding
metal ion binding
GABA receptor binding
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0500013359.hg.1 | ENST00000231504.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013359.hg.1 | ENST00000231504.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013359.hg.1 | ENST00000231504.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013359.hg.1 | ENST00000231504.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013359.hg.1 | ENST00000472253.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013359.hg.1 | ENST00000472253.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013359.hg.1 | ENST00000472253.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013359.hg.1 | ENST00000472253.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013359.hg.1 | ENST00000481195.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013359.hg.1 | ENST00000481195.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013359.hg.1 | ENST00000481195.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013359.hg.1 | ENST00000481195.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013359.hg.1 | ENST00000481195.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013359.hg.1 | ENST00000481195.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013359.hg.1 | ENST00000481195.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013359.hg.1 | ENST00000481195.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013359.hg.1 | ENST00000481195.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013359.hg.1 | ENST00000481195.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013359.hg.1 | ENST00000481195.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013359.hg.1 | ENST00000481195.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013359.hg.1 | ENST00000481195.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013359.hg.1 | ENST00000481195.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013359.hg.1 | ENST00000495833.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013359.hg.1 | ENST00000495833.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013359.hg.1 | ENST00000495833.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013359.hg.1 | ENST00000522385.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013359.hg.1 | ENST00000522385.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013359.hg.1 | ENST00000522385.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013359.hg.1 | ENST00000522385.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013359.hg.1 | ENST00000523082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013359.hg.1 | ENST00000523082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013359.hg.1 | ENST00000523082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013359.hg.1 | NM_002715.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013359.hg.1 | NM_002715.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013359.hg.1 | NM_002715.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013359.hg.1 | NM_002715.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013359.hg.1 | NM_002715.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013359.hg.1 | NM_002715.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013359.hg.1 | NM_002715.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013359.hg.1 | NM_002715.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013359.hg.1 | NM_002715.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013359.hg.1 | NM_002715.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013359.hg.1 | NM_002715.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013359.hg.1 | NM_002715.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013359.hg.1 | NM_002715.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013359.hg.1 | NM_002715.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05024337.hg.1 | TC05003406.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05024337.hg.1 | NM_002715.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05024325.hg.1 | TC05003406.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05024325.hg.1 | ENST00000481195.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05024325.hg.1 | NM_002715.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008688.hg.1 | ENST00000577394.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008688.hg.1 | NR_037434.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05024329.hg.1 | TC05003406.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05024329.hg.1 | TC05003407.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05024329.hg.1 | ENST00000231504.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05024329.hg.1 | ENST00000481195.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05024329.hg.1 | NM_002715.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05024330.hg.1 | TC05003406.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05024330.hg.1 | TC05003407.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05024330.hg.1 | ENST00000231504.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05024330.hg.1 | ENST00000481195.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05024330.hg.1 | ENST00000518409.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05024330.hg.1 | NM_002715.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013848.hg.1 | ENST00000413739.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013848.hg.1 | ENST00000413739.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013848.hg.1 | ENST00000413739.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05024333.hg.1 | TC05003406.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05024333.hg.1 | ENST00000231504.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05024333.hg.1 | ENST00000481195.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05024333.hg.1 | NM_002715.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05024336.hg.1 | TC05003406.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05024336.hg.1 | ENST00000481195.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05024336.hg.1 | NM_002715.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500196647.hg.1 | ENST00000481195.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500196647.hg.1 | NM_002715.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500196647.hg.1 | ENST00000481195.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500196647.hg.1 | NM_002715.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500196647.hg.1 | TC0500013359.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0100013848.hg.1 | ENST00000413739.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013848.hg.1 | ENST00000413739.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013848.hg.1 | ENST00000413739.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500196644.hg.1 | ENST00000231504.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500196644.hg.1 | ENST00000481195.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500196644.hg.1 | NM_002715.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500196644.hg.1 | ENST00000231504.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500196644.hg.1 | ENST00000481195.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500196644.hg.1 | NM_002715.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500196644.hg.1 | TC0500013359.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0500008688.hg.1 | ENST00000577394.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008688.hg.1 | NR_037434.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500196648.hg.1 | NM_002715.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500196648.hg.1 | NM_002715.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500196648.hg.1 | TC0500013359.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0500196638.hg.1 | ENST00000481195.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500196638.hg.1 | ENST00000495833.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500196638.hg.1 | ENST00000522385.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500196638.hg.1 | ENST00000523082.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500196638.hg.1 | NM_002715.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500196638.hg.1 | ENST00000481195.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500196638.hg.1 | ENST00000495833.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500196638.hg.1 | ENST00000522385.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500196638.hg.1 | ENST00000523082.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500196638.hg.1 | NM_002715.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500196638.hg.1 | TC0500013359.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0500196645.hg.1 | ENST00000231504.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500196645.hg.1 | ENST00000481195.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500196645.hg.1 | NM_002715.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500196645.hg.1 | ENST00000231504.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500196645.hg.1 | ENST00000481195.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500196645.hg.1 | NM_002715.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500196645.hg.1 | TC0500013359.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0500196641.hg.1 | ENST00000231504.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500196641.hg.1 | ENST00000481195.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500196641.hg.1 | ENST00000522385.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500196641.hg.1 | ENST00000523082.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500196641.hg.1 | NM_002715.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500196641.hg.1 | ENST00000231504.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500196641.hg.1 | ENST00000481195.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500196641.hg.1 | ENST00000522385.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500196641.hg.1 | ENST00000523082.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500196641.hg.1 | NM_002715.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500196641.hg.1 | TC0500013359.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0500013359.hg.1 | ENST00000231504.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013359.hg.1 | ENST00000231504.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013359.hg.1 | ENST00000231504.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013359.hg.1 | ENST00000231504.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013359.hg.1 | ENST00000472253.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013359.hg.1 | ENST00000472253.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013359.hg.1 | ENST00000472253.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013359.hg.1 | ENST00000472253.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013359.hg.1 | ENST00000481195.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013359.hg.1 | ENST00000481195.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013359.hg.1 | ENST00000481195.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013359.hg.1 | ENST00000481195.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013359.hg.1 | ENST00000481195.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013359.hg.1 | ENST00000481195.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013359.hg.1 | ENST00000481195.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013359.hg.1 | ENST00000481195.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013359.hg.1 | ENST00000481195.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013359.hg.1 | ENST00000481195.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013359.hg.1 | ENST00000481195.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013359.hg.1 | ENST00000481195.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013359.hg.1 | ENST00000481195.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013359.hg.1 | ENST00000481195.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013359.hg.1 | ENST00000495833.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013359.hg.1 | ENST00000495833.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013359.hg.1 | ENST00000495833.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013359.hg.1 | ENST00000522385.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013359.hg.1 | ENST00000522385.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013359.hg.1 | ENST00000522385.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013359.hg.1 | ENST00000522385.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013359.hg.1 | ENST00000523082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013359.hg.1 | ENST00000523082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013359.hg.1 | ENST00000523082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013359.hg.1 | NM_002715.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013359.hg.1 | NM_002715.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013359.hg.1 | NM_002715.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013359.hg.1 | NM_002715.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013359.hg.1 | NM_002715.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013359.hg.1 | NM_002715.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013359.hg.1 | NM_002715.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013359.hg.1 | NM_002715.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013359.hg.1 | NM_002715.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013359.hg.1 | NM_002715.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013359.hg.1 | NM_002715.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013359.hg.1 | NM_002715.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013359.hg.1 | NM_002715.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013359.hg.1 | NM_002715.3 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 101256 | - | - |
ABI Human Genome Survey Microarray Version 2.0 | 188684 | - | - |
