Assay Details
| Gene Symbol: NACC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_052876.3 | NP_443108.1 | 2 - 3 | 1122 | 116 | |
| XM_005259721.3 | XP_005259778.1 | 3 - 4 | 1181 | 116 | ||
| GenBank mRNA | AF395817.1 | - | 2 - 3 | 1073 | 116 | |
| BC055396.1 | - | 3 - 4 | 1056 | 6142288 | 116 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19000236.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19003496.hg.1 | ENST00000292431.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19003496.hg.1 | NM_052876.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19003496.hg.1 | TC19000236.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19003495.hg.1 | ENST00000292431.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19003495.hg.1 | NM_052876.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19003495.hg.1 | TC19000236.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19003493.hg.1 | ENST00000292431.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19003493.hg.1 | NM_052876.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19003493.hg.1 | TC19000236.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19003494.hg.1 | ENST00000292431.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19003494.hg.1 | NM_052876.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19003494.hg.1 | TC19000236.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19003497.hg.1 | ENST00000292431.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19003497.hg.1 | NM_052876.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19003497.hg.1 | TC19000236.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007125.hg.1 | ENST00000292431.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007125.hg.1 | ENST00000292431.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007125.hg.1 | ENST00000292431.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007125.hg.1 | ENST00000292431.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007125.hg.1 | ENST00000292431.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007125.hg.1 | ENST00000292431.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007125.hg.1 | ENST00000292431.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007125.hg.1 | ENST00000292431.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007125.hg.1 | ENST00000292431.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007125.hg.1 | ENST00000292431.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007125.hg.1 | ENST00000292431.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007125.hg.1 | ENST00000292431.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007125.hg.1 | ENST00000585663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007125.hg.1 | ENST00000585663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007125.hg.1 | ENST00000585663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007125.hg.1 | ENST00000586171.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007125.hg.1 | ENST00000586171.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007125.hg.1 | NM_052876.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007125.hg.1 | NM_052876.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007125.hg.1 | NM_052876.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007125.hg.1 | NM_052876.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007125.hg.1 | NM_052876.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007125.hg.1 | NM_052876.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007125.hg.1 | NM_052876.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007125.hg.1 | NM_052876.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007125.hg.1 | NM_052876.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007125.hg.1 | NM_052876.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007125.hg.1 | NM_052876.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007125.hg.1 | NM_052876.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007125.hg.1 | NM_052876.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900151277.hg.1 | ENST00000292431.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900151277.hg.1 | NM_052876.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900151277.hg.1 | TC1900007125.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900151277.hg.1 | ENST00000292431.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900151277.hg.1 | NM_052876.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900151278.hg.1 | ENST00000292431.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900151278.hg.1 | NM_052876.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900151278.hg.1 | TC1900007125.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900151278.hg.1 | ENST00000292431.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900151278.hg.1 | NM_052876.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900151274.hg.1 | ENST00000292431.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900151274.hg.1 | NM_052876.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900151274.hg.1 | TC1900007125.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900151274.hg.1 | ENST00000292431.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900151274.hg.1 | NM_052876.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900151271.hg.1 | NM_052876.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900151271.hg.1 | TC1900007125.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900151271.hg.1 | NM_052876.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900151272.hg.1 | ENST00000292431.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900151272.hg.1 | NM_052876.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900151272.hg.1 | TC1900007125.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900151272.hg.1 | ENST00000292431.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900151272.hg.1 | NM_052876.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900151273.hg.1 | ENST00000292431.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900151273.hg.1 | NM_052876.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900151273.hg.1 | TC1900007125.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900151273.hg.1 | ENST00000292431.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900151273.hg.1 | NM_052876.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900151276.hg.1 | ENST00000292431.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900151276.hg.1 | ENST00000586171.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900151276.hg.1 | NM_052876.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900151276.hg.1 | TC1900007125.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900151276.hg.1 | ENST00000292431.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900151276.hg.1 | ENST00000586171.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900151276.hg.1 | NM_052876.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007125.hg.1 | ENST00000292431.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007125.hg.1 | ENST00000292431.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007125.hg.1 | ENST00000292431.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007125.hg.1 | ENST00000292431.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007125.hg.1 | ENST00000292431.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007125.hg.1 | ENST00000292431.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007125.hg.1 | ENST00000292431.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007125.hg.1 | ENST00000292431.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007125.hg.1 | ENST00000292431.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007125.hg.1 | ENST00000292431.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007125.hg.1 | ENST00000292431.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007125.hg.1 | ENST00000292431.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007125.hg.1 | ENST00000585663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007125.hg.1 | ENST00000585663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007125.hg.1 | ENST00000585663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007125.hg.1 | ENST00000586171.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007125.hg.1 | ENST00000586171.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007125.hg.1 | NM_052876.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007125.hg.1 | NM_052876.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007125.hg.1 | NM_052876.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007125.hg.1 | NM_052876.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007125.hg.1 | NM_052876.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007125.hg.1 | NM_052876.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007125.hg.1 | NM_052876.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007125.hg.1 | NM_052876.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007125.hg.1 | NM_052876.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007125.hg.1 | NM_052876.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007125.hg.1 | NM_052876.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007125.hg.1 | NM_052876.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007125.hg.1 | NM_052876.3 | Refseq |

NACC1
Genomic Map