Assay Details
Gene Symbol: PICALM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008660.2 | NP_001008660.1 | 16 - 17 | 1947 | 148 | |
XM_005274322.2 | XP_005274379.1 | 17 - 18 | 2081 | 148 | ||
XM_005274323.2 | XP_005274380.1 | 17 - 18 | 2066 | 148 | ||
XM_005274326.2 | XP_005274383.1 | 17 - 18 | 2081 | 148 | ||
XM_005274328.2 | XP_005274385.1 | 17 - 18 | 2066 | 148 | ||
XM_005274330.2 | XP_005274387.1 | 16 - 17 | 1931 | 148 | ||
XM_005274332.2 | XP_005274389.1 | 16 - 17 | 1916 | 148 | ||
XM_005274336.2 | XP_005274393.1 | 16 - 17 | 1916 | 148 | ||
XM_006718699.2 | XP_006718762.1 | 17 - 18 | 2060 | 148 | ||
XM_006718700.2 | XP_006718763.1 | 17 - 18 | 2045 | 148 | ||
XM_011545293.1 | XP_011543595.1 | 15 - 16 | 1823 | 148 | ||
XM_017018381.1 | XP_016873870.1 | 17 - 18 | 2045 | 148 | ||
XM_017018388.1 | XP_016873877.1 | 15 - 16 | 1823 | 148 | ||
GenBank mRNA | AB451308.1 | - | 16 - 17 | 148 | ||
AK297062.1 | - | 8 - 9 | 148 | |||
BC064357.1 | - | 16 - 17 | 1909 | 5112878 | 148 | |
BC073961.1 | - | 16 - 17 | 1783 | 6183858 | 148 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
