Assay Details
Gene Symbol: ZNF790 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_206894.3 | NP_996777.2 | 1 - 2 | 49 | 86 | |
GenBank mRNA | AK297474.1 | - | 1 - 2 | 58 | 86 | |
BC057245.1 | - | 1 - 2 | 51 | 6712908 | 86 | |
DA819130.1 | - | 1 - 2 | 47 | 86 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC19001472.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK297474.1,BC057245.1,NM_206894.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR19020019.hg.1 | ENST00000356725.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19020019.hg.1 | NM_206894.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19020019.hg.1 | TC19001472.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000356725.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000356725.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000356725.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000356725.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000356725.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000356725.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000356725.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000356725.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000356725.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000356725.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000525288.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000525288.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000525288.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000525288.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000525288.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000527645.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000527645.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000527645.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000528994.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000528994.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000528994.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000528994.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000528994.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000528994.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000528994.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000586323.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000586323.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000586323.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000586323.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000586323.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000613249.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000613249.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000613249.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000613249.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000613249.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000613249.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000613249.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000613249.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000614179.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000614179.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000614179.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000614179.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000614179.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000614179.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000614179.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000614179.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000615484.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000615484.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000615484.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000615484.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000615484.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000615484.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000615484.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | ENST00000615484.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | NM_001242800.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | NM_001242800.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | NM_001242800.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | NM_001242800.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | NM_001242800.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | NM_001242800.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | NM_001242800.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | NM_001242800.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | NM_001242801.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | NM_001242801.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | NM_001242801.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | NM_001242801.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | NM_001242801.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | NM_001242801.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | NM_001242801.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | NM_001242801.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | NM_001242802.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | NM_001242802.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | NM_001242802.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | NM_001242802.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | NM_001242802.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | NM_001242802.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | NM_001242802.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | NM_001242802.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | NM_206894.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | NM_206894.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | NM_206894.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | NM_206894.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | NM_206894.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | NM_206894.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | NM_206894.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010538.hg.1 | NM_206894.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900186613.hg.1 | ENST00000356725.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900186613.hg.1 | NM_206894.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900186613.hg.1 | TC1900010538.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900186613.hg.1 | ENST00000356725.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900186613.hg.1 | NM_206894.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000356725.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000356725.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000356725.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000356725.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000356725.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000356725.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000356725.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000356725.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000356725.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000356725.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000525288.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000525288.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000525288.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000525288.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000525288.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000527645.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000527645.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000527645.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000528994.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000528994.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000528994.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000528994.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000528994.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000528994.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000528994.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000586323.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000586323.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000586323.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000586323.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000586323.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000613249.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000613249.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000613249.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000613249.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000613249.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000613249.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000613249.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000613249.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000614179.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000614179.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000614179.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000614179.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000614179.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000614179.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000614179.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000614179.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000615484.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000615484.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000615484.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000615484.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000615484.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000615484.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000615484.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | ENST00000615484.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | NM_001242800.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | NM_001242800.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | NM_001242800.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | NM_001242800.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | NM_001242800.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | NM_001242800.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | NM_001242800.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | NM_001242800.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | NM_001242801.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | NM_001242801.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | NM_001242801.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | NM_001242801.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | NM_001242801.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | NM_001242801.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | NM_001242801.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | NM_001242801.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | NM_001242802.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | NM_001242802.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | NM_001242802.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | NM_001242802.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | NM_001242802.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | NM_001242802.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | NM_001242802.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | NM_001242802.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | NM_206894.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | NM_206894.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | NM_206894.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | NM_206894.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | NM_206894.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | NM_206894.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | NM_206894.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010538.hg.1 | NM_206894.3 | Refseq |
