Assay Details
| Gene Symbol: AFG3L1P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_003226.1 | 3 - 4 | 455 | 67 | ||
| NR_003227.1 | 3 - 4 | 382 | 67 | |||
| NR_003228.1 | 3 - 4 | 421 | 67 | |||
| GenBank mRNA | AF329691.1 | - | 2 - 3 | 206 | 67 | |
| AF329693.1 | - | 2 - 3 | 206 | 67 | ||
| AK056488.1 | - | 3 - 4 | 408 | 67 | ||
| AK097044.1 | - | 3 - 4 | 369 | 67 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16000710.hg
Additional Information:
This assay is in the 5'UTR region of the AK056488.1,AK097044.1 transcripts.
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000355531.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000355531.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000355531.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000355531.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000355531.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000355531.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000355531.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000355531.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000355531.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000388970.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000388970.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000388970.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000388970.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000388970.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000388970.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000388970.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000388970.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000388970.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000388970.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000388970.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000388970.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000418696.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000418696.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000418696.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000418696.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000418696.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000418696.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000418696.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000418696.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000418696.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000418696.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000418696.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000421164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000421164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000421164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000421164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000421164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000421164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000421164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000421164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000421164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000421164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000421164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000421164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000421780.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000421780.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000421780.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000421780.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000421780.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000423684.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000423684.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000423684.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000423684.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000423684.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000423684.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000423742.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000423742.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000423742.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000423742.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000427658.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000427658.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000427658.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000427658.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000427658.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000429663.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000429663.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000429663.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000429663.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000431757.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000431757.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000431757.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000431757.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000431757.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000436447.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000436447.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000436447.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000436447.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000436447.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000436447.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000436447.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000436447.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000436447.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000436447.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000436447.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000437774.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000437774.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000437774.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000437774.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000437774.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000437774.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000437774.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000437774.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000437774.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000437774.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000437774.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000437774.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000437774.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000437774.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000450026.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000450026.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000450026.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000450026.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000454997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000454997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000454997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000454997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000454997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000454997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000454997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000454997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000454997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000457926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000457926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000457926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000457926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000457926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000457926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000457926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000457926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000457926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000457926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000458301.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000458301.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000458301.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000458301.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000458301.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000557444.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000557444.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000557444.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000557444.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000557444.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000557444.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000557444.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000557444.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000557444.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000557444.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000557444.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000557444.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000557444.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000557444.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000557444.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000557444.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000557444.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000557444.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000557444.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000557444.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003226.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003226.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003226.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003226.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003226.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003226.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003226.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003226.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003226.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003226.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003226.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003226.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003226.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003226.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003227.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003227.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003227.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003227.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003227.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003227.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003227.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003227.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003227.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003227.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003228.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003228.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003228.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003228.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003228.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003228.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003228.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003228.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003228.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003228.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003228.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003228.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003228.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003228.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003228.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003228.1 | Refseq |

AFG3L1P
Genomic Map