Assay Details
| Gene Symbol: ARHGDIG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001176.3 | NP_001167.2 | 2 - 3 | 331 | 87 | |
| GenBank mRNA | AB593111.1 | - | 2 - 3 | 490 | 87 | |
| AF498928.1 | - | 2 - 3 | 256 | 87 | ||
| BC047699.1 | - | 2 - 3 | 332 | 5581679 | 87 | |
| U82532.1 | - | 2 - 3 | 294 | 87 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16002048.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of cell adhesion
Rho protein signal transduction
regulation of protein localization
positive regulation of GTPase activity
regulation of small GTPase mediated signal transduction
Rho GDP-dissociation inhibitor activity
GTPase activator activity
protein binding
negative regulation of cell adhesion
Rho protein signal transduction
regulation of protein localization
positive regulation of GTPase activity
regulation of small GTPase mediated signal transduction
Rho GDP-dissociation inhibitor activity
GTPase activator activity
protein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR16000176.hg.1 | TC16002048.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16000176.hg.1 | ENST00000219409.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16000176.hg.1 | NM_001176.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011316.hg.1 | ENST00000219409.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011316.hg.1 | ENST00000219409.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011316.hg.1 | ENST00000219409.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011316.hg.1 | ENST00000219409.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011316.hg.1 | ENST00000219409.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011316.hg.1 | ENST00000219409.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011316.hg.1 | ENST00000219409.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011316.hg.1 | ENST00000219409.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011316.hg.1 | ENST00000219409.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011316.hg.1 | ENST00000219409.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011316.hg.1 | ENST00000414650.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011316.hg.1 | ENST00000414650.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011316.hg.1 | ENST00000414650.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011316.hg.1 | ENST00000414650.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011316.hg.1 | ENST00000414650.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011316.hg.1 | ENST00000414650.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011316.hg.1 | ENST00000414650.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011316.hg.1 | ENST00000447871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011316.hg.1 | ENST00000447871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011316.hg.1 | ENST00000447871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011316.hg.1 | ENST00000447871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011316.hg.1 | ENST00000447871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011316.hg.1 | ENST00000447871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011316.hg.1 | ENST00000447871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011316.hg.1 | ENST00000447871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011316.hg.1 | ENST00000477621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011316.hg.1 | ENST00000477621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011316.hg.1 | ENST00000477621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011316.hg.1 | ENST00000477621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011316.hg.1 | ENST00000477621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011316.hg.1 | ENST00000477621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011316.hg.1 | ENST00000477621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011316.hg.1 | ENST00000477621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011316.hg.1 | NM_001176.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011316.hg.1 | NM_001176.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011316.hg.1 | NM_001176.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011316.hg.1 | NM_001176.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011316.hg.1 | NM_001176.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011316.hg.1 | NM_001176.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011316.hg.1 | NM_001176.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011316.hg.1 | NM_001176.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011316.hg.1 | NM_001176.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011316.hg.1 | NM_001176.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16000171.hg.1 | TC16002048.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16000171.hg.1 | ENST00000219409.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16000171.hg.1 | NM_001176.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16000174.hg.1 | TC16002048.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16000174.hg.1 | ENST00000219409.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16000174.hg.1 | NM_001176.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16000169.hg.1 | TC16002048.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16000169.hg.1 | ENST00000219409.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16000169.hg.1 | NM_001176.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011316.hg.1 | ENST00000219409.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011316.hg.1 | ENST00000219409.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011316.hg.1 | ENST00000219409.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011316.hg.1 | ENST00000219409.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011316.hg.1 | ENST00000219409.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011316.hg.1 | ENST00000219409.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011316.hg.1 | ENST00000219409.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011316.hg.1 | ENST00000219409.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011316.hg.1 | ENST00000219409.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011316.hg.1 | ENST00000219409.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011316.hg.1 | ENST00000414650.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011316.hg.1 | ENST00000414650.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011316.hg.1 | ENST00000414650.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011316.hg.1 | ENST00000414650.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011316.hg.1 | ENST00000414650.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011316.hg.1 | ENST00000414650.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011316.hg.1 | ENST00000414650.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011316.hg.1 | ENST00000447871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011316.hg.1 | ENST00000447871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011316.hg.1 | ENST00000447871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011316.hg.1 | ENST00000447871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011316.hg.1 | ENST00000447871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011316.hg.1 | ENST00000447871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011316.hg.1 | ENST00000447871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011316.hg.1 | ENST00000447871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011316.hg.1 | ENST00000477621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011316.hg.1 | ENST00000477621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011316.hg.1 | ENST00000477621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011316.hg.1 | ENST00000477621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011316.hg.1 | ENST00000477621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011316.hg.1 | ENST00000477621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011316.hg.1 | ENST00000477621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011316.hg.1 | ENST00000477621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011316.hg.1 | NM_001176.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011316.hg.1 | NM_001176.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011316.hg.1 | NM_001176.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011316.hg.1 | NM_001176.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011316.hg.1 | NM_001176.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011316.hg.1 | NM_001176.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011316.hg.1 | NM_001176.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011316.hg.1 | NM_001176.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011316.hg.1 | NM_001176.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011316.hg.1 | NM_001176.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600188485.hg.1 | ENST00000219409.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600188485.hg.1 | ENST00000414650.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600188485.hg.1 | ENST00000447871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600188485.hg.1 | ENST00000477621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600188485.hg.1 | NM_001176.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600188485.hg.1 | ENST00000219409.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600188485.hg.1 | ENST00000414650.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600188485.hg.1 | ENST00000447871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600188485.hg.1 | ENST00000477621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600188485.hg.1 | NM_001176.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600188485.hg.1 | TC1600011316.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600188482.hg.1 | ENST00000219409.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600188482.hg.1 | ENST00000447871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600188482.hg.1 | NM_001176.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600188482.hg.1 | ENST00000219409.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600188482.hg.1 | ENST00000447871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600188482.hg.1 | NM_001176.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600188482.hg.1 | TC1600011316.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600188487.hg.1 | ENST00000219409.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600188487.hg.1 | ENST00000414650.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600188487.hg.1 | ENST00000447871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600188487.hg.1 | ENST00000477621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600188487.hg.1 | NM_001176.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600188487.hg.1 | ENST00000219409.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600188487.hg.1 | ENST00000414650.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600188487.hg.1 | ENST00000447871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600188487.hg.1 | ENST00000477621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600188487.hg.1 | NM_001176.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600188487.hg.1 | TC1600011316.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600188481.hg.1 | ENST00000219409.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600188481.hg.1 | ENST00000447871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600188481.hg.1 | NM_001176.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600188481.hg.1 | ENST00000219409.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600188481.hg.1 | ENST00000447871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600188481.hg.1 | NM_001176.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600188481.hg.1 | TC1600011316.hg.1 | Clariom D TC |

ARHGDIG
Genomic Map