Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08005748.hg.1 |
TC08000412.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08005748.hg.1 |
ENST00000307121.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08005748.hg.1 |
ENST00000423902.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08005748.hg.1 |
ENST00000525508.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08005748.hg.1 |
NM_017780.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800153447.hg.1 |
ENST00000423902.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800153447.hg.1 |
ENST00000525508.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800153447.hg.1 |
NM_017780.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800153447.hg.1 |
ENST00000423902.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800153447.hg.1 |
ENST00000525508.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800153447.hg.1 |
NM_017780.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800153447.hg.1 |
TC0800007774.hg.1 |
Clariom D TC |