Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11017905.hg.1 |
ENST00000227756.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11017905.hg.1 |
NM_198516.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11017905.hg.1 |
TC11001422.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010105.hg.1 |
ENST00000227756.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010105.hg.1 |
ENST00000227756.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010105.hg.1 |
ENST00000227756.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010105.hg.1 |
ENST00000227756.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010105.hg.1 |
ENST00000227756.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010105.hg.1 |
ENST00000227756.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010105.hg.1 |
ENST00000227756.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010105.hg.1 |
ENST00000227756.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010105.hg.1 |
ENST00000227756.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010105.hg.1 |
ENST00000227756.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010105.hg.1 |
ENST00000227756.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010105.hg.1 |
ENST00000227756.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010105.hg.1 |
ENST00000227756.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010105.hg.1 |
ENST00000227756.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010105.hg.1 |
ENST00000526064.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010105.hg.1 |
ENST00000526064.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010105.hg.1 |
ENST00000526064.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010105.hg.1 |
ENST00000526064.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010105.hg.1 |
NM_198516.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010105.hg.1 |
NM_198516.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010105.hg.1 |
NM_198516.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010105.hg.1 |
NM_198516.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010105.hg.1 |
NM_198516.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010105.hg.1 |
NM_198516.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010105.hg.1 |
NM_198516.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010105.hg.1 |
NM_198516.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010105.hg.1 |
NM_198516.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010105.hg.1 |
NM_198516.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010105.hg.1 |
NM_198516.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010105.hg.1 |
NM_198516.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010105.hg.1 |
NM_198516.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010105.hg.1 |
NM_198516.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100176735.hg.1 |
ENST00000227756.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100176735.hg.1 |
NM_198516.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100176735.hg.1 |
ENST00000227756.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100176735.hg.1 |
NM_198516.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100176735.hg.1 |
TC1100010105.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100010105.hg.1 |
ENST00000227756.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010105.hg.1 |
ENST00000227756.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010105.hg.1 |
ENST00000227756.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010105.hg.1 |
ENST00000227756.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010105.hg.1 |
ENST00000227756.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010105.hg.1 |
ENST00000227756.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010105.hg.1 |
ENST00000227756.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010105.hg.1 |
ENST00000227756.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010105.hg.1 |
ENST00000227756.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010105.hg.1 |
ENST00000227756.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010105.hg.1 |
ENST00000227756.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010105.hg.1 |
ENST00000227756.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010105.hg.1 |
ENST00000227756.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010105.hg.1 |
ENST00000227756.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010105.hg.1 |
ENST00000526064.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010105.hg.1 |
ENST00000526064.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010105.hg.1 |
ENST00000526064.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010105.hg.1 |
ENST00000526064.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010105.hg.1 |
NM_198516.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010105.hg.1 |
NM_198516.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010105.hg.1 |
NM_198516.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010105.hg.1 |
NM_198516.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010105.hg.1 |
NM_198516.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010105.hg.1 |
NM_198516.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010105.hg.1 |
NM_198516.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010105.hg.1 |
NM_198516.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010105.hg.1 |
NM_198516.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010105.hg.1 |
NM_198516.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010105.hg.1 |
NM_198516.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010105.hg.1 |
NM_198516.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010105.hg.1 |
NM_198516.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010105.hg.1 |
NM_198516.2 |
Refseq |