Assay Details
Gene Symbol: PARVA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018222.4 | NP_060692.2 | 7 - 8 | 889 | 74 | |
XM_005253015.3 | XP_005253072.1 | 7 - 8 | 878 | 74 | ||
GenBank mRNA | AF237771.1 | - | 7 - 8 | 812 | 74 | |
AF325830.1 | - | 7 - 8 | 720 | 74 | ||
AK001655.1 | - | 7 - 8 | 769 | 74 | ||
AK098504.1 | - | 6 - 7 | 813 | 74 | ||
AK304444.1 | - | 7 - 8 | 697 | 74 | ||
BC014535.1 | - | 7 - 8 | 748 | 4065820 | 74 | |
BC016713.1 | - | 7 - 8 | 806 | 4065758 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
sprouting angiogenesis
outflow tract septum morphogenesis
establishment or maintenance of cell polarity
regulation of cell shape
actin cytoskeleton reorganization
heterotypic cell-cell adhesion
substrate adhesion-dependent cell spreading
cilium morphogenesis
actin-mediated cell contraction
smooth muscle cell chemotaxis
actin binding
protein binding
cadherin binding involved in cell-cell adhesion
sprouting angiogenesis
outflow tract septum morphogenesis
establishment or maintenance of cell polarity
regulation of cell shape
actin cytoskeleton reorganization
heterotypic cell-cell adhesion
substrate adhesion-dependent cell spreading
cilium morphogenesis
actin-mediated cell contraction
smooth muscle cell chemotaxis
actin binding
protein binding
cadherin binding involved in cell-cell adhesion
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000526746.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000526746.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000526746.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000526746.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000528916.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000528916.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000528916.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000528916.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000528916.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000528916.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000528916.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000528916.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000528916.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000528916.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000530755.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000530755.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000530755.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000530755.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000530755.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000530755.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000530755.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000533345.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000533345.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000533345.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000533345.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000533345.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000533345.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000533345.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000533345.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000533345.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000533345.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000533345.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000533345.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000533392.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000533392.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000533392.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000533392.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000526746.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000526746.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000526746.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000526746.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000528916.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000528916.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000528916.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000528916.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000528916.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000528916.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000528916.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000528916.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000528916.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000528916.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000530755.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000530755.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000530755.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000530755.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000530755.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000530755.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000530755.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000533345.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000533345.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000533345.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000533345.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000533345.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000533345.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000533345.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000533345.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000533345.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000533345.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000533345.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000533345.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000533392.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000533392.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000533392.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000533392.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
