Assay Details
| Gene Symbol: CRP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC020766.1 | - | 2 - 3 | 305 | 62 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | ENST00000255030.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | ENST00000255030.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | ENST00000255030.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | ENST00000255030.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | ENST00000255030.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | ENST00000255030.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | ENST00000255030.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | ENST00000255030.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | ENST00000255030.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | ENST00000255030.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | ENST00000255030.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | ENST00000255030.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | ENST00000255030.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | ENST00000255030.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | ENST00000255030.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | ENST00000255030.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | ENST00000255030.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | ENST00000368110.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | ENST00000368110.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | ENST00000368110.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | ENST00000368110.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | ENST00000368110.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | ENST00000368110.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | ENST00000368110.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | ENST00000368111.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | ENST00000368111.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | ENST00000368111.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | ENST00000368111.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | ENST00000368111.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | ENST00000368111.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | ENST00000368111.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | ENST00000368111.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | ENST00000368112.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | ENST00000368112.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | ENST00000368112.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | ENST00000368112.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | ENST00000368112.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | ENST00000368112.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | ENST00000368112.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | ENST00000368112.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | ENST00000368112.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | ENST00000437342.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | ENST00000437342.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | ENST00000437342.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | ENST00000437342.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | ENST00000437342.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | ENST00000437342.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | ENST00000437342.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | ENST00000437342.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | ENST00000437342.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | ENST00000437342.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | ENST00000437342.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | ENST00000473196.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | ENST00000473196.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | ENST00000473196.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | ENST00000473196.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | ENST00000473196.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | ENST00000473196.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | ENST00000473196.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | ENST00000473196.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | ENST00000473196.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | ENST00000489317.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | ENST00000489317.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | ENST00000489317.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | NM_000567.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | NM_000567.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | NM_000567.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | NM_000567.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | NM_000567.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | NM_000567.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | NM_000567.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | NM_000567.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | NM_000567.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | NM_000567.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | NM_000567.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | NM_000567.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | NM_000567.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | NM_000567.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | NM_000567.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | NM_000567.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016099.hg.1 | NM_000567.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | ENST00000255030.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | ENST00000255030.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | ENST00000255030.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | ENST00000255030.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | ENST00000255030.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | ENST00000255030.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | ENST00000255030.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | ENST00000255030.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | ENST00000255030.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | ENST00000255030.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | ENST00000255030.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | ENST00000255030.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | ENST00000255030.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | ENST00000255030.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | ENST00000255030.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | ENST00000255030.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | ENST00000255030.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | ENST00000368110.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | ENST00000368110.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | ENST00000368110.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | ENST00000368110.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | ENST00000368110.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | ENST00000368110.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | ENST00000368110.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | ENST00000368111.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | ENST00000368111.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | ENST00000368111.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | ENST00000368111.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | ENST00000368111.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | ENST00000368111.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | ENST00000368111.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | ENST00000368111.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | ENST00000368112.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | ENST00000368112.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | ENST00000368112.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | ENST00000368112.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | ENST00000368112.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | ENST00000368112.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | ENST00000368112.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | ENST00000368112.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | ENST00000368112.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | ENST00000437342.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | ENST00000437342.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | ENST00000437342.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | ENST00000437342.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | ENST00000437342.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | ENST00000437342.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | ENST00000437342.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | ENST00000437342.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | ENST00000437342.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | ENST00000437342.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | ENST00000437342.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | ENST00000473196.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | ENST00000473196.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | ENST00000473196.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | ENST00000473196.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | ENST00000473196.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | ENST00000473196.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | ENST00000473196.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | ENST00000473196.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | ENST00000473196.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | ENST00000489317.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | ENST00000489317.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | ENST00000489317.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | NM_000567.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | NM_000567.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | NM_000567.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | NM_000567.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | NM_000567.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | NM_000567.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | NM_000567.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | NM_000567.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | NM_000567.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | NM_000567.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | NM_000567.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | NM_000567.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | NM_000567.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | NM_000567.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | NM_000567.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | NM_000567.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016099.hg.1 | NM_000567.2 | Refseq |

CRP
Genomic Map