Assay Details
| Gene Symbol: KCNA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204269.1 | NP_001191198.1 | 1 - 2 | 79 | 81 | |
| GenBank mRNA | BC043564.1 | - | 1 - 2 | 79 | 5212949 | 81 |
| BI907383.1 | - | 1 - 2 | 79 | 81 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01002975.hg
Additional Information:
This assay is in the 5'UTR region of the BC043564.1,NM_001204269.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
potassium ion transport
regulation of dopamine secretion
neuronal action potential
sensory perception of pain
optic nerve structural organization
regulation of ion transmembrane transport
regulation of circadian sleep/wake cycle, non-REM sleep
protein homooligomerization
potassium ion transmembrane transport
voltage-gated potassium channel activity
delayed rectifier potassium channel activity
potassium channel activity
protein binding
potassium ion transport
regulation of dopamine secretion
neuronal action potential
sensory perception of pain
optic nerve structural organization
regulation of ion transmembrane transport
regulation of circadian sleep/wake cycle, non-REM sleep
protein homooligomerization
potassium ion transmembrane transport
voltage-gated potassium channel activity
delayed rectifier potassium channel activity
potassium channel activity
protein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01046206.hg.1 | TC01002975.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046206.hg.1 | ENST00000369770.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046206.hg.1 | NM_001204269.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000316361.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000316361.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000316361.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000316361.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000316361.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000316361.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000316361.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000316361.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000369770.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000369770.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000369770.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000369770.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000369770.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000369770.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000369770.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000369770.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000369770.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000485317.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000485317.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000485317.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000485317.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000485317.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000485317.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000485317.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000485317.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000485317.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000485317.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000525120.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000525120.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000525120.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000633222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000633222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000633222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000633222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000633222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000633222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000633222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000633222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000633222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000633222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000633222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000633222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000633222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000633222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000633222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | NM_001204269.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | NM_001204269.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | NM_001204269.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | NM_001204269.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | NM_001204269.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | NM_001204269.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | NM_001204269.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | NM_001204269.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | NM_001204269.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | NM_004974.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | NM_004974.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | NM_004974.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | NM_004974.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | NM_004974.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | NM_004974.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | NM_004974.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | NM_004974.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | NM_004974.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046201.hg.1 | TC01002975.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046201.hg.1 | ENST00000316361.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046201.hg.1 | ENST00000369770.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046201.hg.1 | ENST00000440270.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046201.hg.1 | ENST00000485317.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046201.hg.1 | ENST00000525120.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046201.hg.1 | NM_001204269.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046201.hg.1 | NM_004974.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000316361.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000316361.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000316361.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000316361.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000316361.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000316361.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000316361.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000316361.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000369770.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000369770.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000369770.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000369770.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000369770.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000369770.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000369770.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000369770.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000369770.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000485317.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000485317.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000485317.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000485317.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000485317.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000485317.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000485317.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000485317.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000485317.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000485317.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000525120.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000525120.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000525120.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000633222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000633222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000633222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000633222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000633222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000633222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000633222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000633222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000633222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000633222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000633222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000633222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000633222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000633222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000633222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | NM_001204269.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | NM_001204269.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | NM_001204269.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | NM_001204269.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | NM_001204269.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | NM_001204269.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | NM_001204269.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | NM_001204269.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | NM_001204269.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | NM_004974.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | NM_004974.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | NM_004974.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | NM_004974.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | NM_004974.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | NM_004974.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | NM_004974.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | NM_004974.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | NM_004974.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100213648.hg.1 | ENST00000316361.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213648.hg.1 | ENST00000369770.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213648.hg.1 | ENST00000485317.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213648.hg.1 | ENST00000525120.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213648.hg.1 | ENST00000633222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213648.hg.1 | NM_001204269.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100213648.hg.1 | NM_004974.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100213648.hg.1 | ENST00000316361.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213648.hg.1 | ENST00000369770.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213648.hg.1 | ENST00000485317.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213648.hg.1 | ENST00000525120.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213648.hg.1 | ENST00000633222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213648.hg.1 | NM_001204269.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100213648.hg.1 | NM_004974.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100213648.hg.1 | TC0100015244.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100213653.hg.1 | ENST00000369770.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213653.hg.1 | NM_001204269.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100213653.hg.1 | ENST00000369770.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213653.hg.1 | NM_001204269.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100213653.hg.1 | TC0100015244.hg.1 | Clariom D TC |

KCNA2
Genomic Map