Assay Details
| Gene Symbol: LRRC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011534110.2 | XP_011532412.1 | 1 - 2 | 160 | 122 | |
| GenBank mRNA | BC029118.1 | - | 1 - 2 | 143 | 5183639 | 122 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC029118.1, XM_011534110.2 transcripts.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | ENST00000296144.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | ENST00000296144.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | ENST00000296144.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | ENST00000296144.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | ENST00000296144.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | ENST00000296144.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | ENST00000296144.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | ENST00000296144.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | ENST00000296144.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | ENST00000296144.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | ENST00000296144.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | ENST00000395905.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | ENST00000395905.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | ENST00000395905.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | ENST00000395905.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | ENST00000395905.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | ENST00000395905.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | ENST00000395905.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | ENST00000395905.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | ENST00000395905.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | ENST00000395905.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | ENST00000395905.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | ENST00000395905.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | ENST00000395905.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | ENST00000395905.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | ENST00000469912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | ENST00000469912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | ENST00000469912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | ENST00000496388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | ENST00000496388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | ENST00000496388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | NM_024512.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | NM_024512.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | NM_024512.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | NM_024512.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | NM_024512.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | NM_024512.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | NM_024512.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | NM_024512.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | NM_024512.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | NM_024512.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | NM_024512.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | NM_024512.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | NM_024512.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | NM_024512.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | NM_024512.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300179841.hg.1 | ENST00000296144.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300179841.hg.1 | ENST00000296144.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300179841.hg.1 | TC0300010952.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | ENST00000296144.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | ENST00000296144.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | ENST00000296144.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | ENST00000296144.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | ENST00000296144.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | ENST00000296144.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | ENST00000296144.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | ENST00000296144.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | ENST00000296144.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | ENST00000296144.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | ENST00000296144.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | ENST00000395905.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | ENST00000395905.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | ENST00000395905.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | ENST00000395905.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | ENST00000395905.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | ENST00000395905.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | ENST00000395905.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | ENST00000395905.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | ENST00000395905.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | ENST00000395905.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | ENST00000395905.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | ENST00000395905.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | ENST00000395905.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | ENST00000395905.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | ENST00000469912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | ENST00000469912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | ENST00000469912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | ENST00000496388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | ENST00000496388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | ENST00000496388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | NM_024512.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | NM_024512.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | NM_024512.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | NM_024512.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | NM_024512.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | NM_024512.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | NM_024512.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | NM_024512.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | NM_024512.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | NM_024512.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | NM_024512.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | NM_024512.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | NM_024512.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | NM_024512.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | NM_024512.4 | Refseq |

LRRC2
Genomic Map