Assay Details
Gene Symbol: MORC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085354.2 | NP_001078823.1 | 16 - 17 | 2898 | 61 | |
XM_017029844.1 | XP_016885333.1 | 15 - 16 | 2788 | 61 | ||
GenBank mRNA | AK300305.1 | - | 15 - 16 | 2473 | 61 | |
EF125209.1 | - | 16 - 17 | 2623 | 61 | ||
EF125210.1 | - | 15 - 16 | 2682 | 61 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC0X001257.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR0X016955.hg.1 | TC0X001257.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X016955.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X016955.hg.1 | ENST00000355610.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X016955.hg.1 | ENST00000535534.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X016955.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X016955.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000478924.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000478924.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000478924.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000478924.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000478924.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000478924.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000604604.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | ENST00000604604.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000478924.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000478924.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000478924.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000478924.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000478924.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000478924.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000604604.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | ENST00000604604.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010459.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00171215.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00171215.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00171215.hg.1 | ENST00000478924.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00171215.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00171215.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00171215.hg.1 | ENST00000255495.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00171215.hg.1 | ENST00000355610.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00171215.hg.1 | ENST00000478924.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00171215.hg.1 | NM_001085354.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00171215.hg.1 | NM_024657.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00171215.hg.1 | TC0X00010459.hg.1 | Clariom D TC |
