Assay Details
| Gene Symbol: MORC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085354.2 | NP_001078823.1 | 12 - 13 | 1720 | 93 | |
| NM_024657.4 | NP_078933.3 | 12 - 13 | 1720 | 93 | ||
| XM_005262190.3 | XP_005262247.1 | 11 - 12 | 1610 | 93 | ||
| XM_006724691.2 | XP_006724754.1 | 10 - 11 | 1354 | 93 | ||
| XM_011531027.2 | XP_011529329.1 | 9 - 10 | 1008 | 93 | ||
| XM_017029844.1 | XP_016885333.1 | 11 - 12 | 1610 | 93 | ||
| GenBank mRNA | AK056235.1 | - | 8 - 9 | 932 | 93 | |
| AK300305.1 | - | 11 - 12 | 1295 | 93 | ||
| EF125209.1 | - | 12 - 13 | 1445 | 93 | ||
| EF125210.1 | - | 11 - 12 | 1504 | 93 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

MORC4
Genomic Map