Assay Details
| Gene Symbol: SEMA4C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017789.4 | NP_060259.4 | 3 - 4 | 419 | 73 | |
| XM_006712606.3 | XP_006712669.2 | 3 - 4 | 523 | 73 | ||
| XM_011511378.2 | XP_011509680.1 | 3 - 4 | 527 | 73 | ||
| XM_011511379.2 | XP_011509681.1 | 3 - 4 | 524 | 73 | ||
| XM_011511380.1 | XP_011509682.1 | 3 - 4 | 862 | 73 | ||
| XM_011511381.1 | XP_011509683.1 | 3 - 4 | 496 | 73 | ||
| XM_011511382.2 | XP_011509684.1 | 3 - 4 | 455 | 73 | ||
| XM_011511383.1 | XP_011509685.1 | 3 - 4 | 467 | 73 | ||
| XM_017004393.1 | XP_016859882.1 | 2 - 3 | 949 | 73 | ||
| GenBank mRNA | AB051526.1 | - | 1 - 2 | 649 | 73 | |
| AY358842.1 | - | 3 - 4 | 392 | 73 | ||
| BC109103.1 | - | 3 - 4 | 348 | 40035252 | 73 | |
| BC109104.1 | - | 3 - 4 | 345 | 40035253 | 73 | |
| BI518922.1 | - | 3 - 4 | 420 | 73 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02002113.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
neural crest cell migration
neural tube closure
cell migration in hindbrain
cerebellum development
positive regulation of cell migration
positive regulation of stress-activated MAPK cascade
muscle cell differentiation
negative regulation of axon extension involved in axon guidance
negative chemotaxis
semaphorin-plexin signaling pathway
protein binding
semaphorin receptor binding
neuropilin binding
chemorepellent activity
neural crest cell migration
neural tube closure
cell migration in hindbrain
cerebellum development
positive regulation of cell migration
positive regulation of stress-activated MAPK cascade
muscle cell differentiation
negative regulation of axon extension involved in axon guidance
negative chemotaxis
semaphorin-plexin signaling pathway
protein binding
semaphorin receptor binding
neuropilin binding
chemorepellent activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | ENST00000305476.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | ENST00000305476.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | ENST00000305476.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | ENST00000305476.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | ENST00000305476.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | ENST00000305476.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | ENST00000305476.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | ENST00000305476.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | ENST00000305476.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | ENST00000305476.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | ENST00000305476.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | ENST00000305476.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | ENST00000305476.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | ENST00000305476.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | ENST00000305476.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | ENST00000305476.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | ENST00000305476.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | ENST00000305476.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | ENST00000305476.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | ENST00000305476.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | ENST00000442264.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | ENST00000442264.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | ENST00000442264.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | ENST00000442264.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | ENST00000442264.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | ENST00000442264.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | ENST00000442264.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | ENST00000442264.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | ENST00000449330.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | ENST00000449330.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | ENST00000449330.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | ENST00000449330.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | ENST00000449330.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | ENST00000449330.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | ENST00000467747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | ENST00000467747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | ENST00000467747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | ENST00000467747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | ENST00000467747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | ENST00000467747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | ENST00000467747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | ENST00000474420.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | ENST00000474420.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | ENST00000474420.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | ENST00000474420.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | ENST00000474420.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | ENST00000474420.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | ENST00000474420.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | ENST00000482925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | ENST00000482925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | ENST00000482925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | ENST00000482925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | ENST00000482925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | ENST00000482925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | ENST00000482925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | ENST00000482925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | ENST00000482925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | ENST00000482925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | ENST00000482925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | ENST00000482925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | ENST00000482925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | ENST00000482925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | ENST00000482925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | ENST00000482925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | ENST00000482925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | NM_017789.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | NM_017789.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | NM_017789.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | NM_017789.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | NM_017789.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | NM_017789.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | NM_017789.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | NM_017789.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | NM_017789.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | NM_017789.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | NM_017789.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | NM_017789.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | NM_017789.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | NM_017789.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | NM_017789.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | NM_017789.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | NM_017789.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | NM_017789.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | NM_017789.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | NM_017789.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013541.hg.1 | NM_017789.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02034290.hg.1 | TC02002113.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02034290.hg.1 | ENST00000305476.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02034290.hg.1 | NM_017789.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | ENST00000305476.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | ENST00000305476.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | ENST00000305476.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | ENST00000305476.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | ENST00000305476.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | ENST00000305476.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | ENST00000305476.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | ENST00000305476.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | ENST00000305476.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | ENST00000305476.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | ENST00000305476.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | ENST00000305476.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | ENST00000305476.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | ENST00000305476.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | ENST00000305476.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | ENST00000305476.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | ENST00000305476.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | ENST00000305476.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | ENST00000305476.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | ENST00000305476.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | ENST00000442264.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | ENST00000442264.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | ENST00000442264.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | ENST00000442264.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | ENST00000442264.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | ENST00000442264.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | ENST00000442264.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | ENST00000442264.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | ENST00000449330.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | ENST00000449330.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | ENST00000449330.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | ENST00000449330.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | ENST00000449330.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | ENST00000449330.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | ENST00000467747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | ENST00000467747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | ENST00000467747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | ENST00000467747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | ENST00000467747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | ENST00000467747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | ENST00000467747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | ENST00000474420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | ENST00000474420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | ENST00000474420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | ENST00000474420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | ENST00000474420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | ENST00000474420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | ENST00000474420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | ENST00000482925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | ENST00000482925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | ENST00000482925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | ENST00000482925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | ENST00000482925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | ENST00000482925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | ENST00000482925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | ENST00000482925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | ENST00000482925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | ENST00000482925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | ENST00000482925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | ENST00000482925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | ENST00000482925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | ENST00000482925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | ENST00000482925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | ENST00000482925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | ENST00000482925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | NM_017789.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | NM_017789.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | NM_017789.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | NM_017789.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | NM_017789.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | NM_017789.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | NM_017789.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | NM_017789.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | NM_017789.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | NM_017789.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | NM_017789.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | NM_017789.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | NM_017789.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | NM_017789.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | NM_017789.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | NM_017789.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | NM_017789.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | NM_017789.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | NM_017789.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | NM_017789.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013541.hg.1 | NM_017789.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200199156.hg.1 | ENST00000305476.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200199156.hg.1 | ENST00000442264.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200199156.hg.1 | ENST00000449330.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200199156.hg.1 | ENST00000482925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200199156.hg.1 | NM_017789.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200199156.hg.1 | ENST00000305476.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200199156.hg.1 | ENST00000442264.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200199156.hg.1 | ENST00000449330.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200199156.hg.1 | ENST00000482925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200199156.hg.1 | NM_017789.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200199156.hg.1 | TC0200013541.hg.1 | Clariom D TC |

SEMA4C
Genomic Map