Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03037968.hg.1 |
TC03002152.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03037968.hg.1 |
ENST00000333602.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03037968.hg.1 |
ENST00000381916.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03037968.hg.1 |
ENST00000392400.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03037968.hg.1 |
ENST00000428187.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03037968.hg.1 |
NM_001010938.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03037968.hg.1 |
NM_005781.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300202018.hg.1 |
ENST00000333602.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300202018.hg.1 |
ENST00000381916.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300202018.hg.1 |
ENST00000416152.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300202018.hg.1 |
ENST00000420716.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300202018.hg.1 |
ENST00000428187.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300202018.hg.1 |
NM_001010938.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300202018.hg.1 |
NM_001308046.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300202018.hg.1 |
NM_005781.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300202018.hg.1 |
ENST00000333602.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300202018.hg.1 |
ENST00000381916.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300202018.hg.1 |
ENST00000416152.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300202018.hg.1 |
ENST00000420716.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300202018.hg.1 |
ENST00000428187.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300202018.hg.1 |
NM_001010938.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300202018.hg.1 |
NM_001308046.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300202018.hg.1 |
NM_005781.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300202018.hg.1 |
TC0300013670.hg.1 |
Clariom D TC |