Assay Details
Gene Symbol: ITGA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_003637.4 | NP_003628.2 | 5 - 6 | 651 | 90 | |
XM_005277436.3 | XP_005277493.1 | 4 - 5 | 402 | 90 | ||
XM_017002622.1 | XP_016858111.1 | 3 - 4 | 608 | 90 | ||
XM_017002623.1 | XP_016858112.1 | 3 - 4 | 608 | 90 | ||
XM_017002624.1 | XP_016858113.1 | 3 - 4 | 608 | 90 | ||
XM_017002625.1 | XP_016858114.1 | 3 - 4 | 608 | 90 | ||
XM_017002627.1 | XP_016858116.1 | 3 - 4 | 608 | 90 | ||
GenBank mRNA | AF074015.1 | - | 5 - 6 | 497 | 90 | |
AF112345.1 | - | 5 - 6 | 552 | 90 | ||
AK304183.1 | - | 4 - 5 | 481 | 90 | ||
AK314255.1 | - | 5 - 6 | 500 | 90 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01001095.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000369304.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000369304.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000369304.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000369304.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000369304.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000369304.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000369304.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000369304.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000369304.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000369304.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000369304.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000369304.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000369304.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000369304.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000369304.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000369304.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000369304.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000369304.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000369304.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000369304.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000369304.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000369304.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000369304.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000369304.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000369304.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000369304.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000369304.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000369304.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000369304.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000369304.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000369304.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000369304.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000369304.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000369304.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000369304.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000369304.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000468261.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000468261.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000468261.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000468261.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000468261.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000468261.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000468261.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000468261.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000468261.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000481236.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000481236.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000481236.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000481236.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000539363.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000539363.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000539363.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000539363.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000539363.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000539363.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000539363.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000539363.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000539363.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000539363.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000539363.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000539363.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000539363.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000539363.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000539363.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000539363.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000539363.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000539363.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000539363.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000539363.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000539363.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000539363.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000539363.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000539363.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000539363.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000539363.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000539363.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000539363.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000539363.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000539363.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000539363.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303040.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303040.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303040.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303040.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303040.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303040.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303040.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303040.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303040.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303040.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303040.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303040.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303040.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303040.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303040.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303040.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303040.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303040.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303040.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303040.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303040.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303040.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303040.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303040.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303040.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303040.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303040.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303040.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303040.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303040.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303040.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303040.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303040.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303041.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303041.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303041.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303041.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303041.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303041.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303041.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303041.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303041.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303041.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303041.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303041.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303041.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303041.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303041.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303041.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303041.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303041.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303041.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303041.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303041.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303041.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303041.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303041.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303041.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303041.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303041.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303041.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303041.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303041.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303041.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303041.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_003637.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_003637.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_003637.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_003637.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_003637.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_003637.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_003637.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_003637.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_003637.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_003637.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_003637.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_003637.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_003637.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_003637.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_003637.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_003637.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_003637.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_003637.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_003637.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_003637.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_003637.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_003637.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_003637.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_003637.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_003637.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_003637.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_003637.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_003637.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_003637.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_003637.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_003637.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_003637.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_003637.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_003637.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_003637.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_003637.4 | Refseq |
