Assay Details
| Gene Symbol: RAMP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005854.2 | NP_005845.2 | 2 - 3 | 235 | 86 | |
| GenBank mRNA | AJ001015.1 | - | 2 - 3 | 235 | 86 | |
| AL548945.3 | - | 2 - 3 | 241 | 86 | ||
| AY265458.1 | - | 2 - 3 | 167 | 86 | ||
| BC027975.1 | - | 2 - 3 | 184 | 5229167 | 86 | |
More Information
Set Membership: |
|

RAMP2
Genomic Map